Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 178528237-178528464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTN
Entrez Gene ID
7273
Gene Name
titin
Gene Aliases
CMD1G, CMH9, CMPD4, EOMFC, HMERF, LGMD2J, MYLK5, TMD
UniGene
Hs.134602
Gene Chromosome Location
Chr.2: 178525989-178807423 on Build GRCh38
Transcripts
NM_001256850.1 NM_001267550.2 NM_003319.4 NM_133378.4 NM_133432.3 NM_133437.4 XM_017004819.1 XM_017004820.1 XM_017004821.1 XM_017004822.1 XM_017004823.1 NR_038271.1 NR_038272.1

Gene Symbol
TTN-AS1
Entrez Gene ID
100506866
Gene Name
TTN antisense RNA 1
Gene Aliases
-
UniGene
Hs.710694
Gene Chromosome Location
Chr.2: 178522827-178620217 on Build GRCh38
Transcripts
NM_001256850.1 NM_001267550.2 NM_003319.4 NM_133378.4 NM_133432.3 NM_133437.4 XM_017004819.1 XM_017004820.1 XM_017004821.1 XM_017004822.1 XM_017004823.1 NR_038271.1 NR_038272.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7180341 c.99533C>T p.S33178L Chr.2: 178528414-178528414 on Build GRCh38
3570369 c.80689G>A p.G26897R Chr.2: 178528343-178528343 on Build GRCh38
2895451 c.80718C>T p.F26906F Chr.2: 178528314-178528314 on Build GRCh38
4187377 c.80450A>G p.Y26817C Chr.2: 178528381-178528381 on Build GRCh38
3570359 c.80719C>A p.R26907S Chr.2: 178528313-178528313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759121530 Chr.2: 178528400-178528400 on Build GRCh38
rs770904787 Chr.2: 178528312-178528312 on Build GRCh38
rs903505453 Chr.2: 178528411-178528411 on Build GRCh38
rs753547920 Chr.2: 178528404-178528404 on Build GRCh38
rs397517800 Chr.2: 178528366-178528366 on Build GRCh38
rs112991903 Chr.2: 178528271-178528271 on Build GRCh38
rs757181562 Chr.2: 178528284-178528286 on Build GRCh38
rs932225386 Chr.2: 178528381-178528381 on Build GRCh38
rs794729571 Chr.2: 178528349-178528349 on Build GRCh38
rs142336788 Chr.2: 178528421-178528421 on Build GRCh38
rs755017454 Chr.2: 178528270-178528270 on Build GRCh38
rs749086389 Chr.2: 178528294-178528294 on Build GRCh38
rs748197748 Chr.2: 178528275-178528275 on Build GRCh38
rs774401824 Chr.2: 178528331-178528331 on Build GRCh38
rs760107623 Chr.2: 178528396-178528396 on Build GRCh38
rs759509591 Chr.2: 178528344-178528344 on Build GRCh38
rs770457558 Chr.2: 178528291-178528291 on Build GRCh38
rs1040525694 Chr.2: 178528356-178528356 on Build GRCh38
rs753812861 Chr.2: 178528279-178528281 on Build GRCh38
rs528535076 Chr.2: 178528397-178528397 on Build GRCh38
rs778765016 Chr.2: 178528300-178528300 on Build GRCh38
rs770000719 Chr.2: 178528277-178528277 on Build GRCh38
rs780526209 Chr.2: 178528269-178528269 on Build GRCh38
rs900676745 Chr.2: 178528362-178528362 on Build GRCh38
rs16866378 Chr.2: 178528384-178528384 on Build GRCh38
rs879048717 Chr.2: 178528361-178528361 on Build GRCh38
rs773306658 Chr.2: 178528289-178528289 on Build GRCh38
rs111401822 Chr.2: 178528429-178528429 on Build GRCh38
rs112720067 Chr.2: 178528428-178528428 on Build GRCh38
rs112188483 Chr.2: 178528273-178528273 on Build GRCh38
rs956359578 Chr.2: 178528295-178528295 on Build GRCh38
rs878999828 Chr.2: 178528385-178528385 on Build GRCh38

View Full Product Details