Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205241542-205241787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMCC2
Entrez Gene ID
9911
Gene Name
transmembrane and coiled-coil domain family 2
Gene Aliases
HUCEP11
UniGene
Hs.6360
Gene Chromosome Location
Chr.1: 205227910-205273343 on Build GRCh38
Transcripts
NM_001242925.1 NM_014858.3 XM_005245684.1 XM_005245685.4 XM_005245686.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7313608 c.279G>T p.R93R Chr.1: 205241576-205241576 on Build GRCh38
329020 c.278G>A p.R93Q Chr.1: 205241575-205241575 on Build GRCh38
6124032 c.345C>A p.S115S Chr.1: 205241642-205241642 on Build GRCh38
6802929 c.373C>T p.R125C Chr.1: 205241670-205241670 on Build GRCh38
1267967 c.394G>A p.V132I Chr.1: 205241691-205241691 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138497244 Chr.1: 205241581-205241581 on Build GRCh38
rs757033126 Chr.1: 205241745-205241745 on Build GRCh38
rs746634835 Chr.1: 205241576-205241576 on Build GRCh38
rs967988329 Chr.1: 205241579-205241579 on Build GRCh38
rs968376504 Chr.1: 205241769-205241769 on Build GRCh38
rs140397839 Chr.1: 205241723-205241723 on Build GRCh38
rs146869196 Chr.1: 205241678-205241678 on Build GRCh38
rs775317589 Chr.1: 205241693-205241693 on Build GRCh38
rs767591396 Chr.1: 205241601-205241601 on Build GRCh38
rs1033504397 Chr.1: 205241643-205241643 on Build GRCh38
rs922133780 Chr.1: 205241726-205241726 on Build GRCh38
rs371629736 Chr.1: 205241649-205241649 on Build GRCh38
rs997948416 Chr.1: 205241724-205241724 on Build GRCh38
rs762896631 Chr.1: 205241696-205241696 on Build GRCh38
rs3795557 Chr.1: 205241735-205241735 on Build GRCh38
rs768634241 Chr.1: 205241771-205241771 on Build GRCh38
rs140601524 Chr.1: 205241749-205241749 on Build GRCh38
rs77149546 Chr.1: 205241747-205241747 on Build GRCh38
rs752293588 Chr.1: 205241670-205241670 on Build GRCh38
rs769889858 Chr.1: 205241690-205241690 on Build GRCh38
rs749184990 Chr.1: 205241770-205241770 on Build GRCh38
rs77588763 Chr.1: 205241699-205241699 on Build GRCh38
rs113687212 Chr.1: 205241647-205241647 on Build GRCh38
rs992847761 Chr.1: 205241613-205241613 on Build GRCh38
rs149851869 Chr.1: 205241568-205241568 on Build GRCh38
rs910160085 Chr.1: 205241691-205241691 on Build GRCh38
rs770874236 Chr.1: 205241580-205241580 on Build GRCh38
rs774535336 Chr.1: 205241592-205241592 on Build GRCh38
rs781203767 Chr.1: 205241682-205241682 on Build GRCh38
rs754198015 Chr.1: 205241648-205241648 on Build GRCh38
rs778462308 Chr.1: 205241572-205241572 on Build GRCh38
rs200789897 Chr.1: 205241714-205241714 on Build GRCh38
rs144375621 Chr.1: 205241674-205241674 on Build GRCh38
rs766574643 Chr.1: 205241711-205241711 on Build GRCh38
rs917330139 Chr.1: 205241656-205241656 on Build GRCh38
rs755409965 Chr.1: 205241571-205241571 on Build GRCh38
rs765265891 Chr.1: 205241591-205241591 on Build GRCh38
rs552285360 Chr.1: 205241575-205241575 on Build GRCh38
rs746186087 Chr.1: 205241649-205241651 on Build GRCh38
rs773274832 Chr.1: 205241704-205241704 on Build GRCh38
rs780957575 Chr.1: 205241746-205241746 on Build GRCh38
rs941817824 Chr.1: 205241710-205241710 on Build GRCh38
rs779837835 Chr.1: 205241764-205241764 on Build GRCh38
rs368073852 Chr.1: 205241753-205241753 on Build GRCh38
rs778145198 Chr.1: 205241642-205241642 on Build GRCh38
rs779266840 Chr.1: 205241652-205241652 on Build GRCh38
rs760954041 Chr.1: 205241707-205241707 on Build GRCh38
rs761764527 Chr.1: 205241599-205241599 on Build GRCh38
rs745631951 Chr.1: 205241688-205241688 on Build GRCh38
rs759554538 Chr.1: 205241582-205241582 on Build GRCh38
rs750630944 Chr.1: 205241621-205241621 on Build GRCh38
rs777290569 Chr.1: 205241671-205241671 on Build GRCh38
rs756270240 Chr.1: 205241624-205241624 on Build GRCh38
rs1022119887 Chr.1: 205241765-205241765 on Build GRCh38
rs141994197 Chr.1: 205241574-205241574 on Build GRCh38
rs992038281 Chr.1: 205241654-205241654 on Build GRCh38
rs200429926 Chr.1: 205241727-205241727 on Build GRCh38
rs200967387 Chr.1: 205241741-205241741 on Build GRCh38
rs769495685 Chr.1: 205241763-205241763 on Build GRCh38
rs139793468 Chr.1: 205241732-205241732 on Build GRCh38

View Full Product Details