Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 27311376-27311635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WSB1
Entrez Gene ID
26118
Gene Name
WD repeat and SOCS box containing 1
Gene Aliases
SWIP1, WSB-1
UniGene
Hs.446017
Gene Chromosome Location
Chr.17: 27294080-27313631 on Build GRCh38
Transcripts
NM_015626.8 NM_134265.2 XM_005257963.3 XM_017024433.1 XM_017024434.1 XM_017024435.1 XM_017024436.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6823387 c.1039C>T p.Q347* Chr.17: 27311549-27311549 on Build GRCh38
7403204 c.1053G>C p.L351F Chr.17: 27311563-27311563 on Build GRCh38
1315968 c.1016G>T p.R339I Chr.17: 27311526-27311526 on Build GRCh38
3932519 c.1026G>A p.E342E Chr.17: 27311536-27311536 on Build GRCh38
4264932 c.1106+9A>T p.? Chr.17: 27311625-27311625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757933504 Chr.17: 27311594-27311594 on Build GRCh38
rs759966180 Chr.17: 27311454-27311454 on Build GRCh38
rs756684809 Chr.17: 27311477-27311477 on Build GRCh38
rs371644715 Chr.17: 27311565-27311565 on Build GRCh38
rs746081529 Chr.17: 27311488-27311490 on Build GRCh38
rs371237093 Chr.17: 27311493-27311493 on Build GRCh38
rs549878218 Chr.17: 27311458-27311458 on Build GRCh38
rs777215414 Chr.17: 27311598-27311598 on Build GRCh38
rs766887201 Chr.17: 27311580-27311580 on Build GRCh38
rs533041917 Chr.17: 27311435-27311435 on Build GRCh38
rs762147863 Chr.17: 27311555-27311555 on Build GRCh38
rs903067782 Chr.17: 27311490-27311490 on Build GRCh38
rs745614539 Chr.17: 27311514-27311514 on Build GRCh38
rs774740892 Chr.17: 27311550-27311550 on Build GRCh38
rs767604344 Chr.17: 27311556-27311556 on Build GRCh38
rs764539949 Chr.17: 27311406-27311406 on Build GRCh38
rs746632734 Chr.17: 27311499-27311499 on Build GRCh38
rs546843821 Chr.17: 27311568-27311568 on Build GRCh38
rs564298976 Chr.17: 27311422-27311422 on Build GRCh38
rs149716714 Chr.17: 27311548-27311548 on Build GRCh38
rs561718754 Chr.17: 27311485-27311485 on Build GRCh38
rs73984468 Chr.17: 27311411-27311411 on Build GRCh38
rs746478529 Chr.17: 27311599-27311599 on Build GRCh38
rs754275782 Chr.17: 27311589-27311589 on Build GRCh38
rs756423156 Chr.17: 27311573-27311573 on Build GRCh38
rs139922584 Chr.17: 27311515-27311515 on Build GRCh38
rs763132210 Chr.17: 27311544-27311544 on Build GRCh38
rs538197983 Chr.17: 27311496-27311496 on Build GRCh38
rs780536291 Chr.17: 27311479-27311479 on Build GRCh38
rs771782080 Chr.17: 27311487-27311487 on Build GRCh38
rs199738704 Chr.17: 27311583-27311583 on Build GRCh38
rs776535003 Chr.17: 27311510-27311510 on Build GRCh38
rs770652630 Chr.17: 27311500-27311500 on Build GRCh38
rs775552720 Chr.17: 27311543-27311543 on Build GRCh38
rs1042923305 Chr.17: 27311426-27311426 on Build GRCh38

View Full Product Details