Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46358196-46358451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR2
Entrez Gene ID
729230
Gene Name
C-C motif chemokine receptor 2
Gene Aliases
CC-CKR-2, CCR-2, CCR2A, CCR2B, CD192, CKR2, CKR2A, CKR2B, CMKBR2, MCP-1-R
UniGene
Hs.511794
Gene Chromosome Location
Chr.3: 46353744-46360940 on Build GRCh38
Transcripts
NM_001123041.2 NM_001123396.1 XM_011534069.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3823971 c.813C>A p.F271L Chr.3: 46358340-46358340 on Build GRCh38
1600264 c.697C>T p.R233* Chr.3: 46358224-46358224 on Build GRCh38
1247628 c.694T>A p.C232S Chr.3: 46358221-46358221 on Build GRCh38
345525 c.832G>T p.E278* Chr.3: 46358359-46358359 on Build GRCh38
5427228 c.890A>C p.H297P Chr.3: 46358417-46358417 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934480642 Chr.3: 46358218-46358218 on Build GRCh38
rs765825679 Chr.3: 46358411-46358411 on Build GRCh38
rs371859867 Chr.3: 46358229-46358229 on Build GRCh38
rs201184651 Chr.3: 46358344-46358344 on Build GRCh38
rs866454124 Chr.3: 46358230-46358230 on Build GRCh38
rs1017546867 Chr.3: 46358268-46358268 on Build GRCh38
rs199623670 Chr.3: 46358388-46358388 on Build GRCh38
rs376675621 Chr.3: 46358310-46358310 on Build GRCh38
rs762049291 Chr.3: 46358410-46358410 on Build GRCh38
rs1024376744 Chr.3: 46358371-46358371 on Build GRCh38
rs779353944 Chr.3: 46358259-46358259 on Build GRCh38
rs758746375 Chr.3: 46358343-46358343 on Build GRCh38
rs183084727 Chr.3: 46358387-46358387 on Build GRCh38
rs751438454 Chr.3: 46358356-46358357 on Build GRCh38
rs866870826 Chr.3: 46358363-46358363 on Build GRCh38
rs747673910 Chr.3: 46358346-46358346 on Build GRCh38
rs373111136 Chr.3: 46358376-46358376 on Build GRCh38
rs982279062 Chr.3: 46358355-46358355 on Build GRCh38
rs868667980 Chr.3: 46358418-46358418 on Build GRCh38
rs760787891 Chr.3: 46358266-46358266 on Build GRCh38
rs780628400 Chr.3: 46358345-46358345 on Build GRCh38
rs747196482 Chr.3: 46358400-46358400 on Build GRCh38
rs1799865 Chr.3: 46358307-46358307 on Build GRCh38
rs775750288 Chr.3: 46358393-46358393 on Build GRCh38
rs746514238 Chr.3: 46358219-46358219 on Build GRCh38
rs376895493 Chr.3: 46358236-46358236 on Build GRCh38
rs769023852 Chr.3: 46358401-46358401 on Build GRCh38
rs770681163 Chr.3: 46358391-46358391 on Build GRCh38
rs775636169 Chr.3: 46358265-46358265 on Build GRCh38
rs368302176 Chr.3: 46358220-46358220 on Build GRCh38
rs1026481655 Chr.3: 46358331-46358331 on Build GRCh38
rs539087679 Chr.3: 46358225-46358225 on Build GRCh38
rs766087628 Chr.3: 46358231-46358233 on Build GRCh38
rs762362055 Chr.3: 46358296-46358296 on Build GRCh38
rs769742268 Chr.3: 46358232-46358232 on Build GRCh38
rs553980827 Chr.3: 46358235-46358235 on Build GRCh38
rs199884389 Chr.3: 46358413-46358413 on Build GRCh38
rs772785853 Chr.3: 46358419-46358419 on Build GRCh38
rs772339032 Chr.3: 46358247-46358247 on Build GRCh38
rs200491743 Chr.3: 46358273-46358273 on Build GRCh38
rs754702855 Chr.3: 46358289-46358289 on Build GRCh38
rs776784364 Chr.3: 46358406-46358406 on Build GRCh38

View Full Product Details