Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 135770476-135770705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBXN4
Entrez Gene ID
23190
Gene Name
UBX domain protein 4
Gene Aliases
UBXD2, UBXDC1, erasin
UniGene
Hs.591242
Gene Chromosome Location
Chr.2: 135741619-135785063 on Build GRCh38
Transcripts
NM_014607.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4838917 c.700G>T p.E234* Chr.2: 135770613-135770613 on Build GRCh38
5595531 c.757G>A p.E253K Chr.2: 135770670-135770670 on Build GRCh38
1399559 c.661G>T p.E221* Chr.2: 135770574-135770574 on Build GRCh38
3981748 c.783T>C p.D261D Chr.2: 135770696-135770696 on Build GRCh38
4989344 c.767G>T p.R256I Chr.2: 135770680-135770680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76110853 Chr.2: 135770654-135770654 on Build GRCh38
rs1019495626 Chr.2: 135770524-135770524 on Build GRCh38
rs767963825 Chr.2: 135770526-135770526 on Build GRCh38
rs979494782 Chr.2: 135770604-135770604 on Build GRCh38
rs747973307 Chr.2: 135770617-135770617 on Build GRCh38
rs761086933 Chr.2: 135770557-135770560 on Build GRCh38
rs527887071 Chr.2: 135770538-135770538 on Build GRCh38
rs759800071 Chr.2: 135770520-135770520 on Build GRCh38
rs1054288917 Chr.2: 135770525-135770525 on Build GRCh38
rs778949791 Chr.2: 135770559-135770559 on Build GRCh38
rs528252433 Chr.2: 135770558-135770558 on Build GRCh38
rs1033887880 Chr.2: 135770629-135770629 on Build GRCh38
rs374650338 Chr.2: 135770611-135770611 on Build GRCh38
rs369013309 Chr.2: 135770645-135770645 on Build GRCh38
rs936693775 Chr.2: 135770518-135770518 on Build GRCh38
rs992294387 Chr.2: 135770684-135770684 on Build GRCh38
rs754726853 Chr.2: 135770607-135770607 on Build GRCh38
rs768870192 Chr.2: 135770638-135770640 on Build GRCh38
rs756670979 Chr.2: 135770551-135770551 on Build GRCh38
rs564342732 Chr.2: 135770557-135770557 on Build GRCh38
rs202213104 Chr.2: 135770576-135770576 on Build GRCh38
rs370028479 Chr.2: 135770548-135770548 on Build GRCh38

View Full Product Details