Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49915104-49915337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRYD7
Entrez Gene ID
57213
Gene Name
SPRY domain containing 7
Gene Aliases
C13orf1, CLLD6
UniGene
Hs.44235
Gene Chromosome Location
Chr.13: 49912702-49936501 on Build GRCh38
Transcripts
NM_001127482.2 NM_020456.3 NR_023351.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4400975 c.521A>G p.Q174R Chr.13: 49915133-49915133 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs948045305 Chr.13: 49915290-49915290 on Build GRCh38
rs150672212 Chr.13: 49915300-49915300 on Build GRCh38
rs766813695 Chr.13: 49915190-49915190 on Build GRCh38
rs909866022 Chr.13: 49915279-49915279 on Build GRCh38
rs761191383 Chr.13: 49915171-49915171 on Build GRCh38
rs917559305 Chr.13: 49915166-49915166 on Build GRCh38
rs755291538 Chr.13: 49915196-49915196 on Build GRCh38
rs918477596 Chr.13: 49915268-49915268 on Build GRCh38
rs778682418 Chr.13: 49915200-49915200 on Build GRCh38
rs759634939 Chr.13: 49915242-49915242 on Build GRCh38
rs750460477 Chr.13: 49915143-49915143 on Build GRCh38
rs754231375 Chr.13: 49915191-49915191 on Build GRCh38
rs1037373143 Chr.13: 49915203-49915203 on Build GRCh38
rs369677090 Chr.13: 49915187-49915188 on Build GRCh38
rs949352141 Chr.13: 49915187-49915187 on Build GRCh38
rs767553490 Chr.13: 49915134-49915134 on Build GRCh38

View Full Product Details