Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41936298-41936571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC25
Entrez Gene ID
83538
Gene Name
tetratricopeptide repeat domain 25
Gene Aliases
-
UniGene
Hs.201134
Gene Chromosome Location
Chr.17: 41930605-41966517 on Build GRCh38
Transcripts
NM_031421.3 NR_110662.1 XM_011525323.2 XM_011525324.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3402901 c.429G>A p.G143G Chr.17: 41936504-41936504 on Build GRCh38
4854127 c.407C>T p.S136F Chr.17: 41936482-41936482 on Build GRCh38
6560784 c.398-3C>T p.? Chr.17: 41936470-41936470 on Build GRCh38
6560785 c.398-3C>T p.? Chr.17: 41936470-41936470 on Build GRCh38
4854128 c.407C>T p.S136F Chr.17: 41936482-41936482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370585734 Chr.17: 41936524-41936524 on Build GRCh38
rs73307801 Chr.17: 41936364-41936364 on Build GRCh38
rs202226204 Chr.17: 41936484-41936484 on Build GRCh38
rs782061929 Chr.17: 41936505-41936505 on Build GRCh38
rs782657253 Chr.17: 41936532-41936532 on Build GRCh38
rs562330451 Chr.17: 41936381-41936381 on Build GRCh38
rs782608595 Chr.17: 41936430-41936430 on Build GRCh38
rs782432451 Chr.17: 41936442-41936442 on Build GRCh38
rs782641363 Chr.17: 41936446-41936446 on Build GRCh38
rs375976105 Chr.17: 41936508-41936508 on Build GRCh38
rs782419861 Chr.17: 41936482-41936482 on Build GRCh38
rs782257209 Chr.17: 41936465-41936465 on Build GRCh38
rs782745704 Chr.17: 41936499-41936499 on Build GRCh38
rs782672764 Chr.17: 41936430-41936431 on Build GRCh38
rs146281841 Chr.17: 41936549-41936549 on Build GRCh38
rs782327944 Chr.17: 41936457-41936457 on Build GRCh38
rs532732949 Chr.17: 41936414-41936414 on Build GRCh38
rs781803524 Chr.17: 41936436-41936436 on Build GRCh38
rs997524701 Chr.17: 41936433-41936433 on Build GRCh38
rs782228627 Chr.17: 41936451-41936451 on Build GRCh38
rs376657049 Chr.17: 41936443-41936443 on Build GRCh38
rs782672876 Chr.17: 41936391-41936391 on Build GRCh38
rs552309113 Chr.17: 41936504-41936504 on Build GRCh38
rs372957705 Chr.17: 41936469-41936469 on Build GRCh38
rs886037889 Chr.17: 41936500-41936501 on Build GRCh38
rs782169838 Chr.17: 41936473-41936473 on Build GRCh38
rs567791589 Chr.17: 41936537-41936537 on Build GRCh38
rs531687593 Chr.17: 41936543-41936543 on Build GRCh38
rs782117360 Chr.17: 41936498-41936498 on Build GRCh38
rs187403332 Chr.17: 41936541-41936541 on Build GRCh38

View Full Product Details