Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 172569924-172570179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUCO
Entrez Gene ID
51430
Gene Name
SUN domain containing ossification factor
Gene Aliases
C1orf9, CH1, OPT, SLP1
UniGene
Hs.204559
Gene Chromosome Location
Chr.1: 172532349-172611833 on Build GRCh38
Transcripts
NM_001282750.1 NM_001282751.1 NM_014283.4 NM_016227.3 XM_006711374.2 XM_006711375.2 XM_006711376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5810866 c.973G>T p.E325* Chr.1: 172570163-172570163 on Build GRCh38
6459642 c.1458G>T p.K486N Chr.1: 172570171-172570171 on Build GRCh38
7210093 c.919C>T p.R307* Chr.1: 172570109-172570109 on Build GRCh38
6459640 c.1440A>G p.A480A Chr.1: 172570153-172570153 on Build GRCh38
7392404 c.940G>T p.E314* Chr.1: 172570130-172570130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753571462 Chr.1: 172570059-172570059 on Build GRCh38
rs868477933 Chr.1: 172570152-172570152 on Build GRCh38
rs150365014 Chr.1: 172570060-172570060 on Build GRCh38
rs747135865 Chr.1: 172570084-172570084 on Build GRCh38
rs771126137 Chr.1: 172570090-172570090 on Build GRCh38
rs867753626 Chr.1: 172570100-172570100 on Build GRCh38
rs1012020612 Chr.1: 172570034-172570034 on Build GRCh38
rs138095001 Chr.1: 172570063-172570063 on Build GRCh38
rs544015857 Chr.1: 172570052-172570052 on Build GRCh38
rs934189949 Chr.1: 172570122-172570122 on Build GRCh38
rs1002784498 Chr.1: 172570006-172570006 on Build GRCh38
rs372728522 Chr.1: 172570110-172570110 on Build GRCh38
rs748538305 Chr.1: 172570026-172570026 on Build GRCh38
rs367980125 Chr.1: 172570153-172570153 on Build GRCh38
rs34888249 Chr.1: 172570091-172570091 on Build GRCh38
rs367881307 Chr.1: 172570031-172570031 on Build GRCh38
rs368774692 Chr.1: 172570107-172570107 on Build GRCh38
rs767086208 Chr.1: 172570051-172570051 on Build GRCh38
rs777795358 Chr.1: 172570082-172570082 on Build GRCh38
rs375313397 Chr.1: 172570120-172570120 on Build GRCh38
rs758842214 Chr.1: 172570033-172570033 on Build GRCh38
rs577574808 Chr.1: 172570021-172570024 on Build GRCh38
rs368426579 Chr.1: 172570068-172570068 on Build GRCh38
rs778589551 Chr.1: 172570061-172570061 on Build GRCh38
rs765656842 Chr.1: 172570026-172570026 on Build GRCh38
rs772031767 Chr.1: 172570081-172570081 on Build GRCh38
rs534863341 Chr.1: 172570053-172570053 on Build GRCh38
rs756434070 Chr.1: 172570053-172570061 on Build GRCh38
rs775740222 Chr.1: 172570132-172570132 on Build GRCh38
rs140541870 Chr.1: 172570123-172570123 on Build GRCh38

View Full Product Details