Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19542340-19542565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC47A1
Entrez Gene ID
55244
Gene Name
solute carrier family 47 member 1
Gene Aliases
MATE1
UniGene
Hs.232054
Gene Chromosome Location
Chr.17: 19533854-19579033 on Build GRCh38
Transcripts
NM_018242.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
373592 c.210G>T p.E70D Chr.17: 19542467-19542467 on Build GRCh38
3515049 c.204G>A p.K68K Chr.17: 19542461-19542461 on Build GRCh38
4790442 c.228G>T p.L76L Chr.17: 19542485-19542485 on Build GRCh38
4929689 c.221T>G p.V74G Chr.17: 19542478-19542478 on Build GRCh38
236861 c.224C>T p.T75M Chr.17: 19542481-19542481 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746202120 Chr.17: 19542464-19542464 on Build GRCh38
rs752630607 Chr.17: 19542431-19542431 on Build GRCh38
rs760362357 Chr.17: 19542491-19542491 on Build GRCh38
rs988969121 Chr.17: 19542380-19542380 on Build GRCh38
rs776052635 Chr.17: 19542477-19542477 on Build GRCh38
rs753368724 Chr.17: 19542421-19542421 on Build GRCh38
rs202002655 Chr.17: 19542492-19542492 on Build GRCh38
rs754857969 Chr.17: 19542417-19542417 on Build GRCh38
rs548651981 Chr.17: 19542433-19542433 on Build GRCh38
rs747199363 Chr.17: 19542437-19542437 on Build GRCh38
rs202140517 Chr.17: 19542456-19542456 on Build GRCh38
rs200313255 Chr.17: 19542498-19542498 on Build GRCh38
rs569046401 Chr.17: 19542391-19542391 on Build GRCh38
rs143217368 Chr.17: 19542481-19542481 on Build GRCh38
rs538185995 Chr.17: 19542404-19542404 on Build GRCh38
rs770193866 Chr.17: 19542470-19542470 on Build GRCh38
rs754397495 Chr.17: 19542472-19542473 on Build GRCh38
rs368070813 Chr.17: 19542519-19542519 on Build GRCh38
rs780731946 Chr.17: 19542485-19542494 on Build GRCh38
rs370179846 Chr.17: 19542434-19542434 on Build GRCh38
rs77630697 Chr.17: 19542448-19542448 on Build GRCh38
rs562942627 Chr.17: 19542385-19542385 on Build GRCh38
rs940407693 Chr.17: 19542495-19542495 on Build GRCh38
rs76043271 Chr.17: 19542513-19542513 on Build GRCh38
rs991252777 Chr.17: 19542372-19542372 on Build GRCh38
rs529272683 Chr.17: 19542365-19542365 on Build GRCh38
rs776468116 Chr.17: 19542493-19542493 on Build GRCh38
rs374411316 Chr.17: 19542482-19542482 on Build GRCh38
rs759388828 Chr.17: 19542494-19542494 on Build GRCh38
rs770318633 Chr.17: 19542364-19542364 on Build GRCh38
rs199554387 Chr.17: 19542375-19542375 on Build GRCh38
rs935897192 Chr.17: 19542445-19542445 on Build GRCh38
rs752618560 Chr.17: 19542499-19542499 on Build GRCh38
rs761363688 Chr.17: 19542373-19542373 on Build GRCh38
rs767003150 Chr.17: 19542374-19542374 on Build GRCh38
rs764189066 Chr.17: 19542514-19542514 on Build GRCh38

View Full Product Details