Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134138852-134139125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCF7
Entrez Gene ID
6932
Gene Name
transcription factor 7 (T-cell specific, HMG-box)
Gene Aliases
TCF-1
UniGene
Hs.573153
Gene Chromosome Location
Chr.5: 134114703-134148229 on Build GRCh38
Transcripts
NM_001134851.2 NM_003202.3 NM_201632.3 NM_201634.3 NM_213648.3 NR_033449.1 XM_006714678.3 XM_006714679.3 XM_006714680.3 XM_006714682.2 XM_006714684.2 XM_006714685.3 XM_006714686.3 XM_006714687.3 XM_011543604.2 XM_011543606.1 XM_011543607.2 XM_011543608.2 XM_011543609.2 XM_011543610.2 XM_011543611.2 XM_011543612.2 XM_011543613.2 XM_017009789.1 XM_017009790.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6798495 c.608T>C p.M203T Chr.5: 134139011-134139011 on Build GRCh38
6252512 c.635+3G>T p.? Chr.5: 134139041-134139041 on Build GRCh38
6798497 c.608T>C p.M203T Chr.5: 134139011-134139011 on Build GRCh38
6252510 c.290+3G>T p.? Chr.5: 134139041-134139041 on Build GRCh38
6252513 c.635+3G>T p.? Chr.5: 134139041-134139041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753097928 Chr.5: 134138911-134138911 on Build GRCh38
rs763337009 Chr.5: 134139041-134139041 on Build GRCh38
rs1043782130 Chr.5: 134138972-134138972 on Build GRCh38
rs995230358 Chr.5: 134138899-134138899 on Build GRCh38
rs370732060 Chr.5: 134138901-134138901 on Build GRCh38
rs960972588 Chr.5: 134138934-134138934 on Build GRCh38
rs148020244 Chr.5: 134139010-134139010 on Build GRCh38
rs751761444 Chr.5: 134138940-134138940 on Build GRCh38
rs865908238 Chr.5: 134138943-134138943 on Build GRCh38
rs770330559 Chr.5: 134138927-134138927 on Build GRCh38
rs748492408 Chr.5: 134138925-134138925 on Build GRCh38
rs992767878 Chr.5: 134138938-134138938 on Build GRCh38
rs1052374924 Chr.5: 134139020-134139020 on Build GRCh38
rs755492591 Chr.5: 134138903-134138903 on Build GRCh38
rs769721881 Chr.5: 134138956-134138956 on Build GRCh38
rs774002943 Chr.5: 134138984-134138984 on Build GRCh38
rs370601770 Chr.5: 134138948-134138948 on Build GRCh38
rs376552460 Chr.5: 134138926-134138926 on Build GRCh38
rs758703202 Chr.5: 134138923-134138923 on Build GRCh38
rs768363117 Chr.5: 134138976-134138976 on Build GRCh38
rs367674461 Chr.5: 134139045-134139045 on Build GRCh38
rs756203006 Chr.5: 134139077-134139077 on Build GRCh38
rs753259781 Chr.5: 134139035-134139035 on Build GRCh38
rs999116866 Chr.5: 134139021-134139021 on Build GRCh38
rs749057251 Chr.5: 134138974-134138974 on Build GRCh38
rs925299106 Chr.5: 134138981-134138981 on Build GRCh38
rs760317594 Chr.5: 134139025-134139025 on Build GRCh38
rs781463420 Chr.5: 134139063-134139063 on Build GRCh38
rs911033326 Chr.5: 134139069-134139069 on Build GRCh38
rs765318406 Chr.5: 134138936-134138937 on Build GRCh38
rs201667020 Chr.5: 134139054-134139054 on Build GRCh38
rs144497620 Chr.5: 134138904-134138904 on Build GRCh38
rs562655081 Chr.5: 134138986-134138986 on Build GRCh38
rs780028603 Chr.5: 134138959-134138959 on Build GRCh38
rs757392380 Chr.5: 134139058-134139058 on Build GRCh38
rs757313138 Chr.5: 134138941-134138941 on Build GRCh38
rs752683741 Chr.5: 134139030-134139031 on Build GRCh38
rs749201324 Chr.5: 134139089-134139089 on Build GRCh38
rs750492374 Chr.5: 134139070-134139070 on Build GRCh38
rs373694202 Chr.5: 134138953-134138953 on Build GRCh38
rs545262466 Chr.5: 134139028-134139028 on Build GRCh38
rs891896 Chr.5: 134138900-134138900 on Build GRCh38
rs779922519 Chr.5: 134139088-134139088 on Build GRCh38
rs935358237 Chr.5: 134139013-134139013 on Build GRCh38
rs771888210 Chr.5: 134138988-134138988 on Build GRCh38
rs964176938 Chr.5: 134138898-134138898 on Build GRCh38
rs995679294 Chr.5: 134139075-134139075 on Build GRCh38

View Full Product Details