Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 35369241-35369502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF396
Entrez Gene ID
252884
Gene Name
zinc finger protein 396
Gene Aliases
ZSCAN14
UniGene
Hs.351005
Gene Chromosome Location
Chr.18: 35366697-35377337 on Build GRCh38
Transcripts
NM_001322286.1 NM_001322290.1 NM_145756.2 XM_006722432.3 XM_017025687.1 XM_017025688.1 XM_017025689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1480311 c.880C>G p.L294V Chr.18: 35369343-35369343 on Build GRCh38
4869359 c.869G>A p.R290Q Chr.18: 35369354-35369354 on Build GRCh38
1388571 c.877A>T p.I293F Chr.18: 35369346-35369346 on Build GRCh38
1680336 c.753G>T p.Q251H Chr.18: 35369470-35369470 on Build GRCh38
3525626 c.867C>G p.S289R Chr.18: 35369356-35369356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375219933 Chr.18: 35369340-35369340 on Build GRCh38
rs557349808 Chr.18: 35369457-35369457 on Build GRCh38
rs751142511 Chr.18: 35369449-35369449 on Build GRCh38
rs116404576 Chr.18: 35369350-35369350 on Build GRCh38
rs780973619 Chr.18: 35369360-35369360 on Build GRCh38
rs769759761 Chr.18: 35369376-35369376 on Build GRCh38
rs374004274 Chr.18: 35369460-35369460 on Build GRCh38
rs905416291 Chr.18: 35369432-35369432 on Build GRCh38
rs867241216 Chr.18: 35369290-35369290 on Build GRCh38
rs746772415 Chr.18: 35369278-35369278 on Build GRCh38
rs139312528 Chr.18: 35369288-35369288 on Build GRCh38
rs777040842 Chr.18: 35369396-35369396 on Build GRCh38
rs1029091625 Chr.18: 35369474-35369474 on Build GRCh38
rs753172094 Chr.18: 35369411-35369411 on Build GRCh38
rs755905627 Chr.18: 35369475-35369475 on Build GRCh38
rs149796653 Chr.18: 35369382-35369382 on Build GRCh38
rs760992408 Chr.18: 35369387-35369387 on Build GRCh38
rs774488647 Chr.18: 35369303-35369303 on Build GRCh38
rs745720795 Chr.18: 35369370-35369370 on Build GRCh38
rs116074936 Chr.18: 35369276-35369276 on Build GRCh38
rs745879283 Chr.18: 35369354-35369354 on Build GRCh38
rs762395918 Chr.18: 35369431-35369431 on Build GRCh38
rs200991704 Chr.18: 35369475-35369476 on Build GRCh38
rs761080609 Chr.18: 35369328-35369328 on Build GRCh38
rs949401279 Chr.18: 35369327-35369327 on Build GRCh38
rs766873525 Chr.18: 35369330-35369330 on Build GRCh38
rs566238013 Chr.18: 35369320-35369320 on Build GRCh38
rs770977648 Chr.18: 35369331-35369331 on Build GRCh38
rs759866047 Chr.18: 35369407-35369407 on Build GRCh38
rs535584134 Chr.18: 35369294-35369294 on Build GRCh38
rs765742311 Chr.18: 35369410-35369410 on Build GRCh38
rs757883580 Chr.18: 35369349-35369349 on Build GRCh38
rs374149724 Chr.18: 35369390-35369390 on Build GRCh38
rs145740578 Chr.18: 35369442-35369442 on Build GRCh38
rs115597469 Chr.18: 35369318-35369318 on Build GRCh38
rs867840508 Chr.18: 35369279-35369279 on Build GRCh38
rs779907741 Chr.18: 35369377-35369377 on Build GRCh38
rs762302049 Chr.18: 35369403-35369404 on Build GRCh38
rs200984340 Chr.18: 35369381-35369381 on Build GRCh38
rs369731655 Chr.18: 35369355-35369355 on Build GRCh38
rs756901477 Chr.18: 35369456-35369456 on Build GRCh38
rs112874582 Chr.18: 35369336-35369336 on Build GRCh38
rs777224773 Chr.18: 35369376-35369380 on Build GRCh38
rs761858803 Chr.18: 35369306-35369306 on Build GRCh38
rs773518822 Chr.18: 35369386-35369386 on Build GRCh38
rs535231827 Chr.18: 35369335-35369335 on Build GRCh38
rs192743555 Chr.18: 35369289-35369289 on Build GRCh38

View Full Product Details