Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240507421-240507694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKMY1
Entrez Gene ID
51281
Gene Name
ankyrin repeat and MYND domain containing 1
Gene Aliases
ZMYND13
UniGene
Hs.656615
Gene Chromosome Location
Chr.2: 240468632-240561110 on Build GRCh38
Transcripts
NM_001282771.1 NM_001282780.1 NM_001282781.1 NM_001308375.1 NM_016552.3 NM_017844.3 XM_011511300.2 XM_011511301.1 XM_011511304.2 XM_011511305.2 XM_011511309.2 XM_011511313.2 XM_011511317.1 XM_011511318.1 XM_011511322.1 XM_017004271.1 XM_017004272.1 XM_017004273.1 XM_017004274.1 XM_017004275.1 XM_017004276.1 XM_017004277.1 XM_017004278.1 XM_017004279.1 XM_017004280.1 XM_017004281.1 XM_017004282.1 XM_017004283.1 XM_017004284.1 XM_017004285.1 XM_017004286.1 XM_017004287.1 XM_017004288.1 XM_017004289.1 XM_017004290.1 XM_017004291.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5652285 c.2222A>G p.Y741C Chr.2: 240507597-240507597 on Build GRCh38
2909482 c.2163C>T p.P721P Chr.2: 240507656-240507656 on Build GRCh38
3372705 c.2166C>A p.N722K Chr.2: 240507653-240507653 on Build GRCh38
4092837 c.2151G>T p.Q717H Chr.2: 240507668-240507668 on Build GRCh38
1591999 c.1587+1G>A p.? Chr.2: 240507559-240507559 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886223319 Chr.2: 240507650-240507650 on Build GRCh38
rs553748723 Chr.2: 240507580-240507580 on Build GRCh38
rs534823319 Chr.2: 240507458-240507458 on Build GRCh38
rs565095200 Chr.2: 240507595-240507595 on Build GRCh38
rs1013236731 Chr.2: 240507671-240507671 on Build GRCh38
rs574767219 Chr.2: 240507513-240507513 on Build GRCh38
rs774924188 Chr.2: 240507628-240507628 on Build GRCh38
rs988862679 Chr.2: 240507489-240507489 on Build GRCh38
rs1057290881 Chr.2: 240507631-240507631 on Build GRCh38
rs772930819 Chr.2: 240507603-240507603 on Build GRCh38
rs372488280 Chr.2: 240507544-240507544 on Build GRCh38
rs767983882 Chr.2: 240507562-240507562 on Build GRCh38
rs199542386 Chr.2: 240507522-240507522 on Build GRCh38
rs74388234 Chr.2: 240507535-240507535 on Build GRCh38
rs761145647 Chr.2: 240507657-240507657 on Build GRCh38
rs553314793 Chr.2: 240507461-240507461 on Build GRCh38
rs757775323 Chr.2: 240507556-240507556 on Build GRCh38
rs956243874 Chr.2: 240507462-240507462 on Build GRCh38
rs774168264 Chr.2: 240507645-240507645 on Build GRCh38
rs368319634 Chr.2: 240507542-240507542 on Build GRCh38
rs754057963 Chr.2: 240507673-240507673 on Build GRCh38
rs144808565 Chr.2: 240507620-240507620 on Build GRCh38
rs975034506 Chr.2: 240507494-240507494 on Build GRCh38
rs769346058 Chr.2: 240507623-240507623 on Build GRCh38
rs766030181 Chr.2: 240507530-240507530 on Build GRCh38
rs148825327 Chr.2: 240507570-240507570 on Build GRCh38
rs570582700 Chr.2: 240507445-240507445 on Build GRCh38
rs749278929 Chr.2: 240507577-240507577 on Build GRCh38
rs766900255 Chr.2: 240507659-240507659 on Build GRCh38
rs200108704 Chr.2: 240507536-240507536 on Build GRCh38
rs762355889 Chr.2: 240507636-240507636 on Build GRCh38
rs772054061 Chr.2: 240507601-240507601 on Build GRCh38
rs746809100 Chr.2: 240507604-240507604 on Build GRCh38
rs138163943 Chr.2: 240507617-240507617 on Build GRCh38
rs770534971 Chr.2: 240507635-240507635 on Build GRCh38
rs778898662 Chr.2: 240507593-240507593 on Build GRCh38
rs201767407 Chr.2: 240507569-240507569 on Build GRCh38
rs750777569 Chr.2: 240507656-240507656 on Build GRCh38
rs759154622 Chr.2: 240507621-240507621 on Build GRCh38
rs748798576 Chr.2: 240507521-240507544 on Build GRCh38
rs752079082 Chr.2: 240507551-240507551 on Build GRCh38
rs200515527 Chr.2: 240507653-240507653 on Build GRCh38

View Full Product Details