Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57573479-57573732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF416
Entrez Gene ID
55659
Gene Name
zinc finger protein 416
Gene Aliases
-
UniGene
Hs.247711
Gene Chromosome Location
Chr.19: 57571015-57578938 on Build GRCh38
Transcripts
NM_017879.1 XM_005259051.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3971311 c.402G>A p.A134A Chr.19: 57573502-57573502 on Build GRCh38
1002339 c.257_259delAAG p.E86delE Chr.19: 57573645-57573647 on Build GRCh38
7229324 c.342C>T p.T114T Chr.19: 57573562-57573562 on Build GRCh38
1002338 c.291T>C p.S97S Chr.19: 57573613-57573613 on Build GRCh38
5447345 c.367C>T p.P123S Chr.19: 57573537-57573537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201862818 Chr.19: 57573672-57573672 on Build GRCh38
rs149811743 Chr.19: 57573507-57573507 on Build GRCh38
rs201684897 Chr.19: 57573599-57573599 on Build GRCh38
rs199964547 Chr.19: 57573676-57573676 on Build GRCh38
rs754413304 Chr.19: 57573645-57573647 on Build GRCh38
rs548420526 Chr.19: 57573569-57573569 on Build GRCh38
rs768600581 Chr.19: 57573517-57573517 on Build GRCh38
rs144609015 Chr.19: 57573636-57573636 on Build GRCh38
rs567563930 Chr.19: 57573596-57573596 on Build GRCh38
rs112485279 Chr.19: 57573543-57573543 on Build GRCh38
rs371604477 Chr.19: 57573653-57573653 on Build GRCh38
rs769429263 Chr.19: 57573618-57573618 on Build GRCh38
rs756473214 Chr.19: 57573707-57573707 on Build GRCh38
rs199591686 Chr.19: 57573544-57573544 on Build GRCh38
rs751636394 Chr.19: 57573568-57573568 on Build GRCh38
rs3746222 Chr.19: 57573562-57573562 on Build GRCh38
rs781384909 Chr.19: 57573572-57573572 on Build GRCh38
rs747971810 Chr.19: 57573594-57573594 on Build GRCh38
rs894843886 Chr.19: 57573523-57573523 on Build GRCh38
rs750464688 Chr.19: 57573703-57573703 on Build GRCh38
rs1000993940 Chr.19: 57573504-57573504 on Build GRCh38
rs747111555 Chr.19: 57573502-57573502 on Build GRCh38
rs761123564 Chr.19: 57573624-57573629 on Build GRCh38
rs750699866 Chr.19: 57573579-57573579 on Build GRCh38
rs766852706 Chr.19: 57573634-57573640 on Build GRCh38
rs762181513 Chr.19: 57573684-57573684 on Build GRCh38
rs145751549 Chr.19: 57573515-57573515 on Build GRCh38
rs772664523 Chr.19: 57573540-57573540 on Build GRCh38
rs141067282 Chr.19: 57573604-57573604 on Build GRCh38
rs369230430 Chr.19: 57573530-57573530 on Build GRCh38
rs763433657 Chr.19: 57573561-57573561 on Build GRCh38
rs139688579 Chr.19: 57573503-57573503 on Build GRCh38
rs771977347 Chr.19: 57573533-57573533 on Build GRCh38
rs755011562 Chr.19: 57573696-57573696 on Build GRCh38
rs756348744 Chr.19: 57573592-57573592 on Build GRCh38
rs1015145693 Chr.19: 57573637-57573637 on Build GRCh38
rs746881058 Chr.19: 57573607-57573607 on Build GRCh38
rs775888413 Chr.19: 57573648-57573648 on Build GRCh38
rs367843112 Chr.19: 57573610-57573610 on Build GRCh38
rs767721772 Chr.19: 57573685-57573685 on Build GRCh38
rs146524018 Chr.19: 57573598-57573598 on Build GRCh38
rs752991103 Chr.19: 57573552-57573552 on Build GRCh38
rs763351066 Chr.19: 57573652-57573652 on Build GRCh38
rs765600382 Chr.19: 57573550-57573550 on Build GRCh38
rs759141149 Chr.19: 57573545-57573545 on Build GRCh38
rs752142634 Chr.19: 57573673-57573673 on Build GRCh38
rs773598032 Chr.19: 57573620-57573621 on Build GRCh38
rs778858372 Chr.19: 57573593-57573593 on Build GRCh38
rs138547423 Chr.19: 57573664-57573664 on Build GRCh38
rs774236162 Chr.19: 57573521-57573521 on Build GRCh38
rs551351673 Chr.19: 57573688-57573688 on Build GRCh38
rs748107994 Chr.19: 57573527-57573527 on Build GRCh38
rs532195166 Chr.19: 57573501-57573501 on Build GRCh38

View Full Product Details