Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57750722-57750995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF776
Entrez Gene ID
284309
Gene Name
zinc finger protein 776
Gene Aliases
-
UniGene
Hs.720254
Gene Chromosome Location
Chr.19: 57746796-57758159 on Build GRCh38
Transcripts
NM_173632.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7156413 c.152C>G p.S51C Chr.19: 57750903-57750903 on Build GRCh38
1193867 c.66C>G p.N22K Chr.19: 57750817-57750817 on Build GRCh38
6152565 c.34G>T p.G12C Chr.19: 57750785-57750785 on Build GRCh38
7156412 c.26C>G p.S9C Chr.19: 57750903-57750903 on Build GRCh38
7313944 c.118C>A p.H40N Chr.19: 57750869-57750869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753362404 Chr.19: 57750836-57750836 on Build GRCh38
rs776661070 Chr.19: 57750779-57750779 on Build GRCh38
rs773072194 Chr.19: 57750908-57750908 on Build GRCh38
rs202242198 Chr.19: 57750920-57750920 on Build GRCh38
rs149749249 Chr.19: 57750823-57750823 on Build GRCh38
rs778057426 Chr.19: 57750870-57750870 on Build GRCh38
rs768841531 Chr.19: 57750754-57750754 on Build GRCh38
rs374595080 Chr.19: 57750896-57750896 on Build GRCh38
rs746030880 Chr.19: 57750894-57750894 on Build GRCh38
rs762812689 Chr.19: 57750780-57750780 on Build GRCh38
rs773402292 Chr.19: 57750771-57750771 on Build GRCh38
rs772675553 Chr.19: 57750861-57750861 on Build GRCh38
rs757102087 Chr.19: 57750948-57750949 on Build GRCh38
rs776945062 Chr.19: 57750884-57750884 on Build GRCh38
rs767057189 Chr.19: 57750809-57750809 on Build GRCh38
rs368451355 Chr.19: 57750947-57750947 on Build GRCh38
rs772298958 Chr.19: 57750767-57750767 on Build GRCh38
rs201176749 Chr.19: 57750775-57750775 on Build GRCh38
rs781145645 Chr.19: 57750959-57750960 on Build GRCh38
rs748713222 Chr.19: 57750851-57750851 on Build GRCh38
rs896694225 Chr.19: 57750855-57750855 on Build GRCh38
rs767425021 Chr.19: 57750903-57750903 on Build GRCh38
rs191494220 Chr.19: 57750952-57750952 on Build GRCh38
rs751522645 Chr.19: 57750804-57750804 on Build GRCh38
rs371005770 Chr.19: 57750768-57750768 on Build GRCh38
rs779096807 Chr.19: 57750848-57750848 on Build GRCh38
rs1013845744 Chr.19: 57750857-57750857 on Build GRCh38
rs767265639 Chr.19: 57750795-57750795 on Build GRCh38
rs886428197 Chr.19: 57750928-57750928 on Build GRCh38
rs58446072 Chr.19: 57750814-57750814 on Build GRCh38
rs761810375 Chr.19: 57750897-57750897 on Build GRCh38
rs760233378 Chr.19: 57750943-57750943 on Build GRCh38
rs561800300 Chr.19: 57750853-57750853 on Build GRCh38
rs761718865 Chr.19: 57750806-57750806 on Build GRCh38
rs748623335 Chr.19: 57750760-57750760 on Build GRCh38
rs574431727 Chr.19: 57750752-57750752 on Build GRCh38
rs763757762 Chr.19: 57750799-57750799 on Build GRCh38
rs754563190 Chr.19: 57750954-57750954 on Build GRCh38
rs755399169 Chr.19: 57750846-57750846 on Build GRCh38
rs770850761 Chr.19: 57750883-57750883 on Build GRCh38
rs761017271 Chr.19: 57750772-57750772 on Build GRCh38
rs779477227 Chr.19: 57750756-57750756 on Build GRCh38
rs749926373 Chr.19: 57750813-57750813 on Build GRCh38
rs747100372 Chr.19: 57750878-57750878 on Build GRCh38
rs753030745 Chr.19: 57750845-57750848 on Build GRCh38

View Full Product Details