Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150364046-150364319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCOF1
Entrez Gene ID
6949
Gene Name
treacle ribosome biogenesis factor 1
Gene Aliases
MFD1, TCS, TCS1, treacle
UniGene
Hs.519672
Gene Chromosome Location
Chr.5: 150357639-150400308 on Build GRCh38
Transcripts
NM_000356.3 NM_001008657.2 NM_001135243.1 NM_001135244.1 NM_001135245.1 NM_001195141.1 XM_005268502.3 XM_005268503.3 XM_005268504.3 XM_005268505.3 XM_005268506.3 XM_005268507.3 XM_005268508.3 XM_005268509.3 XM_011537678.2 XM_017009792.1 XM_017009793.1 XM_017009794.1 XM_017009795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6586472 c.238G>C p.D80H Chr.5: 150364186-150364186 on Build GRCh38
4141565 c.261C>T p.S87S Chr.5: 150364209-150364209 on Build GRCh38
4732815 c.263C>T p.S88L Chr.5: 150364211-150364211 on Build GRCh38
3919367 c.265G>A p.E89K Chr.5: 150364213-150364213 on Build GRCh38
5660951 c.207G>T p.A69A Chr.5: 150364155-150364155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757177854 Chr.5: 150364282-150364282 on Build GRCh38
rs755843802 Chr.5: 150364162-150364162 on Build GRCh38
rs527952644 Chr.5: 150364271-150364271 on Build GRCh38
rs762986105 Chr.5: 150364241-150364241 on Build GRCh38
rs765428099 Chr.5: 150364236-150364236 on Build GRCh38
rs147288650 Chr.5: 150364240-150364241 on Build GRCh38
rs761961716 Chr.5: 150364235-150364235 on Build GRCh38
rs556259715 Chr.5: 150364093-150364093 on Build GRCh38
rs758996100 Chr.5: 150364286-150364286 on Build GRCh38
rs373774150 Chr.5: 150364088-150364088 on Build GRCh38
rs777365006 Chr.5: 150364259-150364259 on Build GRCh38
rs775836475 Chr.5: 150364211-150364211 on Build GRCh38
rs574602226 Chr.5: 150364130-150364130 on Build GRCh38
rs772865214 Chr.5: 150364200-150364200 on Build GRCh38
rs777649173 Chr.5: 150364164-150364164 on Build GRCh38
rs894890059 Chr.5: 150364281-150364281 on Build GRCh38
rs946305689 Chr.5: 150364155-150364155 on Build GRCh38
rs753699722 Chr.5: 150364267-150364267 on Build GRCh38
rs985336128 Chr.5: 150364245-150364245 on Build GRCh38
rs779645531 Chr.5: 150364291-150364291 on Build GRCh38
rs200561413 Chr.5: 150364237-150364237 on Build GRCh38
rs765231530 Chr.5: 150364101-150364101 on Build GRCh38
rs371609378 Chr.5: 150364136-150364136 on Build GRCh38
rs757501632 Chr.5: 150364092-150364092 on Build GRCh38
rs745562523 Chr.5: 150364284-150364284 on Build GRCh38
rs112332762 Chr.5: 150364243-150364243 on Build GRCh38
rs748855952 Chr.5: 150364169-150364169 on Build GRCh38
rs778705409 Chr.5: 150364283-150364283 on Build GRCh38
rs367964727 Chr.5: 150364135-150364135 on Build GRCh38
rs145365677 Chr.5: 150364178-150364178 on Build GRCh38
rs752532261 Chr.5: 150364242-150364242 on Build GRCh38
rs760564202 Chr.5: 150364212-150364212 on Build GRCh38
rs764567260 Chr.5: 150364078-150364078 on Build GRCh38
rs748159754 Chr.5: 150364158-150364158 on Build GRCh38
rs771777470 Chr.5: 150364203-150364203 on Build GRCh38
rs745360259 Chr.5: 150364201-150364201 on Build GRCh38
rs777057497 Chr.5: 150364218-150364218 on Build GRCh38
rs773941712 Chr.5: 150364191-150364191 on Build GRCh38

View Full Product Details