Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 24688466-24688691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP12A
Entrez Gene ID
479
Gene Name
ATPase H+/K+ transporting non-gastric alpha2 subunit
Gene Aliases
#945, ATP1AL1, HK
UniGene
Hs.147111
Gene Chromosome Location
Chr.13: 24680411-24711785 on Build GRCh38
Transcripts
NM_001185085.1 NM_001676.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
946162 c.406A>G p.S136G Chr.13: 24688496-24688496 on Build GRCh38
5716240 c.386C>A p.A129E Chr.13: 24688476-24688476 on Build GRCh38
946163 c.432+1G>A p.? Chr.13: 24688523-24688523 on Build GRCh38
6607207 c.409G>A p.D137N Chr.13: 24688499-24688499 on Build GRCh38
6607203 c.432+1G>A p.? Chr.13: 24688523-24688523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1011688290 Chr.13: 24688618-24688618 on Build GRCh38
rs199568379 Chr.13: 24688522-24688522 on Build GRCh38
rs773913245 Chr.13: 24688510-24688510 on Build GRCh38
rs1021858169 Chr.13: 24688647-24688647 on Build GRCh38
rs946297535 Chr.13: 24688661-24688661 on Build GRCh38
rs768131390 Chr.13: 24688507-24688507 on Build GRCh38
rs540957583 Chr.13: 24688640-24688640 on Build GRCh38
rs139532001 Chr.13: 24688494-24688494 on Build GRCh38
rs942798341 Chr.13: 24688557-24688557 on Build GRCh38
rs1054446719 Chr.13: 24688592-24688592 on Build GRCh38
rs999285351 Chr.13: 24688543-24688543 on Build GRCh38
rs35582198 Chr.13: 24688619-24688620 on Build GRCh38
rs752782562 Chr.13: 24688563-24688563 on Build GRCh38
rs369759214 Chr.13: 24688498-24688498 on Build GRCh38
rs765244203 Chr.13: 24688550-24688550 on Build GRCh38
rs753967715 Chr.13: 24688535-24688535 on Build GRCh38
rs758543220 Chr.13: 24688571-24688571 on Build GRCh38
rs755253723 Chr.13: 24688536-24688536 on Build GRCh38
rs893166744 Chr.13: 24688597-24688597 on Build GRCh38
rs578256204 Chr.13: 24688604-24688604 on Build GRCh38
rs766808530 Chr.13: 24688523-24688523 on Build GRCh38

View Full Product Details