Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20999920-21000185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THAP7
Entrez Gene ID
80764
Gene Name
THAP domain containing 7
Gene Aliases
-
UniGene
Hs.512756
Gene Chromosome Location
Chr.22: 20999772-21002115 on Build GRCh38
Transcripts
NM_001008695.1 NM_030573.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
444743 c.832G>C p.E278Q Chr.22: 20999978-20999978 on Build GRCh38
4102841 c.710C>T p.A237V Chr.22: 21000100-21000100 on Build GRCh38
6801057 c.848A>G p.K283R Chr.22: 20999962-20999962 on Build GRCh38
4594333 c.791G>A p.R264Q Chr.22: 21000019-21000019 on Build GRCh38
6801058 c.725G>A p.R242Q Chr.22: 21000085-21000085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750988756 Chr.22: 20999954-20999968 on Build GRCh38
rs570644625 Chr.22: 21000122-21000122 on Build GRCh38
rs766726678 Chr.22: 20999992-20999995 on Build GRCh38
rs747717595 Chr.22: 21000005-21000005 on Build GRCh38
rs777017103 Chr.22: 21000019-21000019 on Build GRCh38
rs766442228 Chr.22: 21000130-21000130 on Build GRCh38
rs756191065 Chr.22: 21000044-21000044 on Build GRCh38
rs200305591 Chr.22: 20999953-20999953 on Build GRCh38
rs747696489 Chr.22: 21000064-21000064 on Build GRCh38
rs898562372 Chr.22: 20999972-20999972 on Build GRCh38
rs771742388 Chr.22: 21000067-21000067 on Build GRCh38
rs752818661 Chr.22: 21000030-21000030 on Build GRCh38
rs371544505 Chr.22: 21000045-21000045 on Build GRCh38
rs752699415 Chr.22: 20999969-20999969 on Build GRCh38
rs561512265 Chr.22: 21000010-21000010 on Build GRCh38
rs777069968 Chr.22: 21000082-21000082 on Build GRCh38
rs772774591 Chr.22: 21000075-21000075 on Build GRCh38
rs778659846 Chr.22: 21000059-21000059 on Build GRCh38
rs746488268 Chr.22: 21000077-21000077 on Build GRCh38
rs757392040 Chr.22: 21000126-21000126 on Build GRCh38
rs781225001 Chr.22: 20999968-20999968 on Build GRCh38
rs113988439 Chr.22: 21000027-21000027 on Build GRCh38
rs376353818 Chr.22: 20999975-20999975 on Build GRCh38
rs746541643 Chr.22: 21000149-21000149 on Build GRCh38
rs755256671 Chr.22: 20999940-20999940 on Build GRCh38
rs937652834 Chr.22: 21000142-21000142 on Build GRCh38
rs749092358 Chr.22: 20999986-20999986 on Build GRCh38
rs201383966 Chr.22: 21000022-21000022 on Build GRCh38
rs954008883 Chr.22: 21000003-21000003 on Build GRCh38
rs531259542 Chr.22: 20999959-20999959 on Build GRCh38
rs979135123 Chr.22: 21000132-21000132 on Build GRCh38
rs539728839 Chr.22: 21000147-21000147 on Build GRCh38
rs368620190 Chr.22: 21000031-21000031 on Build GRCh38
rs764027670 Chr.22: 21000032-21000032 on Build GRCh38
rs745738023 Chr.22: 20999974-20999974 on Build GRCh38
rs374500 Chr.22: 21000138-21000138 on Build GRCh38
rs758024851 Chr.22: 21000146-21000146 on Build GRCh38
rs778630760 Chr.22: 21000144-21000144 on Build GRCh38
rs375087817 Chr.22: 21000086-21000086 on Build GRCh38
rs753787848 Chr.22: 21000137-21000137 on Build GRCh38
rs766866705 Chr.22: 21000011-21000011 on Build GRCh38
rs189794457 Chr.22: 21000021-21000021 on Build GRCh38
rs898641077 Chr.22: 21000161-21000161 on Build GRCh38
rs761108606 Chr.22: 20999989-20999989 on Build GRCh38
rs752530771 Chr.22: 21000145-21000145 on Build GRCh38
rs781665327 Chr.22: 21000160-21000160 on Build GRCh38
rs754681452 Chr.22: 21000053-21000053 on Build GRCh38
rs773783187 Chr.22: 21000009-21000009 on Build GRCh38
rs759973415 Chr.22: 21000085-21000085 on Build GRCh38
rs764375464 Chr.22: 21000105-21000105 on Build GRCh38
rs370872545 Chr.22: 21000156-21000156 on Build GRCh38
rs773854185 Chr.22: 21000004-21000004 on Build GRCh38
rs780055919 Chr.22: 20999984-20999984 on Build GRCh38
rs767392211 Chr.22: 20999957-20999957 on Build GRCh38
rs751679779 Chr.22: 21000042-21000042 on Build GRCh38
rs376570367 Chr.22: 21000051-21000051 on Build GRCh38
rs756255895 Chr.22: 21000078-21000078 on Build GRCh38
rs920695997 Chr.22: 21000102-21000102 on Build GRCh38
rs1031869800 Chr.22: 21000048-21000048 on Build GRCh38
rs754815287 Chr.22: 21000143-21000143 on Build GRCh38
rs771416808 Chr.22: 21000008-21000008 on Build GRCh38
rs778969857 Chr.22: 20999942-20999942 on Build GRCh38
rs371740159 Chr.22: 21000108-21000108 on Build GRCh38
rs369415723 Chr.22: 21000093-21000093 on Build GRCh38
rs138377774 Chr.22: 20999992-20999992 on Build GRCh38

View Full Product Details