Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 14105525-14105798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF519
Entrez Gene ID
162655
Gene Name
zinc finger protein 519
Gene Aliases
HsT2362
UniGene
Hs.352635
Gene Chromosome Location
Chr.18: 14071230-14132494 on Build GRCh38
Transcripts
NM_145287.3 NR_033354.1 XM_011525605.2 XM_017025562.1 XM_017025563.1 XM_017025564.1 XM_017025565.1 XM_017025566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6488515 c.941A>T p.H314L Chr.18: 14105599-14105599 on Build GRCh38
986781 c.935G>T p.R312I Chr.18: 14105605-14105605 on Build GRCh38
5652345 c.803A>T p.K268I Chr.18: 14105737-14105737 on Build GRCh38
986782 c.760G>T p.G254* Chr.18: 14105780-14105780 on Build GRCh38
5772479 c.844C>A p.H282N Chr.18: 14105696-14105696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760849034 Chr.18: 14105640-14105640 on Build GRCh38
rs970915816 Chr.18: 14105712-14105712 on Build GRCh38
rs756251203 Chr.18: 14105717-14105718 on Build GRCh38
rs1024472976 Chr.18: 14105702-14105702 on Build GRCh38
rs755191489 Chr.18: 14105656-14105656 on Build GRCh38
rs150326170 Chr.18: 14105666-14105666 on Build GRCh38
rs757003662 Chr.18: 14105690-14105690 on Build GRCh38
rs773786403 Chr.18: 14105624-14105624 on Build GRCh38
rs761331609 Chr.18: 14105740-14105740 on Build GRCh38
rs137978357 Chr.18: 14105709-14105709 on Build GRCh38
rs757058920 Chr.18: 14105762-14105762 on Build GRCh38
rs767359477 Chr.18: 14105764-14105764 on Build GRCh38
rs576575185 Chr.18: 14105731-14105731 on Build GRCh38
rs768242734 Chr.18: 14105718-14105718 on Build GRCh38
rs769230666 Chr.18: 14105556-14105556 on Build GRCh38
rs753947046 Chr.18: 14105653-14105653 on Build GRCh38
rs768162018 Chr.18: 14105611-14105611 on Build GRCh38
rs552977206 Chr.18: 14105584-14105584 on Build GRCh38
rs558634760 Chr.18: 14105739-14105739 on Build GRCh38
rs748801426 Chr.18: 14105710-14105710 on Build GRCh38
rs777950304 Chr.18: 14105759-14105765 on Build GRCh38
rs188498928 Chr.18: 14105585-14105585 on Build GRCh38
rs775140206 Chr.18: 14105563-14105563 on Build GRCh38
rs759608302 Chr.18: 14105748-14105748 on Build GRCh38
rs754269472 Chr.18: 14105594-14105594 on Build GRCh38
rs753001461 Chr.18: 14105573-14105573 on Build GRCh38
rs772505762 Chr.18: 14105738-14105738 on Build GRCh38
rs1013460311 Chr.18: 14105614-14105614 on Build GRCh38
rs376906490 Chr.18: 14105555-14105555 on Build GRCh38
rs751331291 Chr.18: 14105669-14105669 on Build GRCh38
rs9303799 Chr.18: 14105754-14105754 on Build GRCh38
rs143678021 Chr.18: 14105745-14105745 on Build GRCh38
rs879100146 Chr.18: 14105586-14105586 on Build GRCh38
rs752990600 Chr.18: 14105714-14105714 on Build GRCh38
rs779253854 Chr.18: 14105571-14105571 on Build GRCh38
rs766961540 Chr.18: 14105741-14105741 on Build GRCh38
rs139075088 Chr.18: 14105704-14105704 on Build GRCh38
rs752900622 Chr.18: 14105761-14105761 on Build GRCh38
rs781114696 Chr.18: 14105703-14105703 on Build GRCh38
rs756035448 Chr.18: 14105708-14105708 on Build GRCh38
rs748955821 Chr.18: 14105570-14105570 on Build GRCh38
rs758927535 Chr.18: 14105577-14105577 on Build GRCh38
rs537188224 Chr.18: 14105732-14105732 on Build GRCh38
rs766623188 Chr.18: 14105641-14105641 on Build GRCh38
rs554892340 Chr.18: 14105726-14105726 on Build GRCh38
rs878970327 Chr.18: 14105618-14105618 on Build GRCh38

View Full Product Details