Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 150409657-150409928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSC22D2
Entrez Gene ID
9819
Gene Name
TSC22 domain family member 2
Gene Aliases
TILZ4a, TILZ4b, TILZ4c
UniGene
Hs.722470
Gene Chromosome Location
Chr.3: 150408268-150460119 on Build GRCh38
Transcripts
NM_001303264.1 NM_014779.3 NR_130136.1 XM_011513337.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7155039 c.495C>T p.I165I Chr.3: 150409845-150409845 on Build GRCh38
3940342 c.455C>G p.S152C Chr.3: 150409805-150409805 on Build GRCh38
3846498 c.492G>A p.V164V Chr.3: 150409842-150409842 on Build GRCh38
7155038 c.487C>T p.R163C Chr.3: 150409837-150409837 on Build GRCh38
356511 c.335G>T p.S112I Chr.3: 150409685-150409685 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745408935 Chr.3: 150409704-150409704 on Build GRCh38
rs780083649 Chr.3: 150409679-150409679 on Build GRCh38
rs970620265 Chr.3: 150409787-150409787 on Build GRCh38
rs138286637 Chr.3: 150409803-150409803 on Build GRCh38
rs569868270 Chr.3: 150409730-150409730 on Build GRCh38
rs190464574 Chr.3: 150409857-150409857 on Build GRCh38
rs943245165 Chr.3: 150409738-150409738 on Build GRCh38
rs61734435 Chr.3: 150409702-150409702 on Build GRCh38
rs759574717 Chr.3: 150409805-150409805 on Build GRCh38
rs753196895 Chr.3: 150409811-150409811 on Build GRCh38
rs369100145 Chr.3: 150409782-150409782 on Build GRCh38
rs770624577 Chr.3: 150409701-150409701 on Build GRCh38
rs866702333 Chr.3: 150409708-150409708 on Build GRCh38
rs930391602 Chr.3: 150409705-150409705 on Build GRCh38
rs370012280 Chr.3: 150409687-150409687 on Build GRCh38
rs771761089 Chr.3: 150409709-150409709 on Build GRCh38
rs773051287 Chr.3: 150409788-150409788 on Build GRCh38
rs780721626 Chr.3: 150409860-150409860 on Build GRCh38
rs760761149 Chr.3: 150409716-150409716 on Build GRCh38
rs1013143182 Chr.3: 150409872-150409872 on Build GRCh38
rs745548860 Chr.3: 150409755-150409755 on Build GRCh38
rs531373445 Chr.3: 150409740-150409740 on Build GRCh38
rs781616768 Chr.3: 150409745-150409745 on Build GRCh38
rs756616791 Chr.3: 150409814-150409814 on Build GRCh38
rs761788392 Chr.3: 150409783-150409783 on Build GRCh38
rs150454925 Chr.3: 150409789-150409789 on Build GRCh38
rs561609411 Chr.3: 150409750-150409750 on Build GRCh38
rs893185681 Chr.3: 150409775-150409775 on Build GRCh38
rs781590505 Chr.3: 150409691-150409691 on Build GRCh38
rs748097941 Chr.3: 150409877-150409877 on Build GRCh38
rs746461403 Chr.3: 150409770-150409770 on Build GRCh38
rs764433242 Chr.3: 150409817-150409817 on Build GRCh38
rs940622633 Chr.3: 150409682-150409682 on Build GRCh38
rs971734843 Chr.3: 150409677-150409677 on Build GRCh38
rs775110864 Chr.3: 150409710-150409710 on Build GRCh38
rs769753574 Chr.3: 150409887-150409887 on Build GRCh38
rs758138770 Chr.3: 150409847-150409847 on Build GRCh38
rs1012075352 Chr.3: 150409780-150409780 on Build GRCh38
rs200087702 Chr.3: 150409858-150409858 on Build GRCh38
rs755470637 Chr.3: 150409688-150409688 on Build GRCh38
rs779769949 Chr.3: 150409762-150409762 on Build GRCh38
rs61734436 Chr.3: 150409752-150409752 on Build GRCh38
rs144183125 Chr.3: 150409759-150409759 on Build GRCh38
rs766527760 Chr.3: 150409801-150409801 on Build GRCh38
rs768743840 Chr.3: 150409779-150409779 on Build GRCh38
rs778181419 Chr.3: 150409753-150409753 on Build GRCh38
rs779812397 Chr.3: 150409848-150409848 on Build GRCh38
rs745904760 Chr.3: 150409741-150409741 on Build GRCh38
rs765518874 Chr.3: 150409737-150409737 on Build GRCh38
rs761474177 Chr.3: 150409736-150409736 on Build GRCh38
rs748399998 Chr.3: 150409698-150409698 on Build GRCh38
rs768703026 Chr.3: 150409723-150409723 on Build GRCh38
rs778393786 Chr.3: 150409703-150409703 on Build GRCh38
rs373246794 Chr.3: 150409830-150409830 on Build GRCh38
rs541627096 Chr.3: 150409742-150409742 on Build GRCh38

View Full Product Details