Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70146459-70146720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYSND1
Entrez Gene ID
219743
Gene Name
trypsin domain containing 1
Gene Aliases
NET41
UniGene
Hs.533655
Gene Chromosome Location
Chr.10: 70137981-70146740 on Build GRCh38
Transcripts
NM_001040273.2 NM_173555.3 NR_073580.1 NR_073581.1 NR_073582.1 NR_073591.1 NR_073592.1 NR_073593.1 NR_073594.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6418632 c.68G>A p.R23Q Chr.10: 70146519-70146519 on Build GRCh38
5622091 c.35C>T p.A12V Chr.10: 70146552-70146552 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs898740166 Chr.10: 70146687-70146687 on Build GRCh38
rs770971544 Chr.10: 70146599-70146599 on Build GRCh38
rs549665753 Chr.10: 70146586-70146586 on Build GRCh38
rs1041144512 Chr.10: 70146696-70146696 on Build GRCh38
rs890903337 Chr.10: 70146669-70146669 on Build GRCh38
rs375511768 Chr.10: 70146551-70146551 on Build GRCh38
rs775335996 Chr.10: 70146498-70146498 on Build GRCh38
rs927978727 Chr.10: 70146603-70146603 on Build GRCh38
rs551874180 Chr.10: 70146511-70146511 on Build GRCh38
rs753631832 Chr.10: 70146513-70146513 on Build GRCh38
rs776498674 Chr.10: 70146621-70146621 on Build GRCh38
rs773518819 Chr.10: 70146484-70146484 on Build GRCh38
rs760645918 Chr.10: 70146506-70146506 on Build GRCh38
rs995739321 Chr.10: 70146688-70146688 on Build GRCh38
rs762974915 Chr.10: 70146489-70146489 on Build GRCh38
rs750104168 Chr.10: 70146636-70146636 on Build GRCh38
rs766450887 Chr.10: 70146494-70146494 on Build GRCh38
rs748545640 Chr.10: 70146572-70146572 on Build GRCh38
rs182789121 Chr.10: 70146663-70146663 on Build GRCh38
rs1050806203 Chr.10: 70146660-70146660 on Build GRCh38
rs1048308175 Chr.10: 70146655-70146655 on Build GRCh38
rs762933946 Chr.10: 70146615-70146617 on Build GRCh38
rs369734572 Chr.10: 70146481-70146481 on Build GRCh38
rs779604352 Chr.10: 70146550-70146550 on Build GRCh38
rs781741817 Chr.10: 70146561-70146561 on Build GRCh38
rs768591017 Chr.10: 70146619-70146619 on Build GRCh38
rs761640891 Chr.10: 70146629-70146629 on Build GRCh38
rs780853620 Chr.10: 70146526-70146526 on Build GRCh38
rs527409953 Chr.10: 70146497-70146497 on Build GRCh38
rs981463644 Chr.10: 70146593-70146593 on Build GRCh38
rs969378680 Chr.10: 70146594-70146594 on Build GRCh38
rs913802116 Chr.10: 70146634-70146634 on Build GRCh38
rs537662173 Chr.10: 70146542-70146542 on Build GRCh38
rs921070251 Chr.10: 70146646-70146646 on Build GRCh38
rs931120095 Chr.10: 70146633-70146633 on Build GRCh38
rs71472947 Chr.10: 70146646-70146647 on Build GRCh38
rs750050724 Chr.10: 70146531-70146531 on Build GRCh38
rs761534631 Chr.10: 70146523-70146523 on Build GRCh38
rs996069844 Chr.10: 70146545-70146545 on Build GRCh38
rs999277056 Chr.10: 70146512-70146512 on Build GRCh38
rs942620187 Chr.10: 70146673-70146673 on Build GRCh38
rs778179980 Chr.10: 70146589-70146589 on Build GRCh38
rs1023857832 Chr.10: 70146566-70146566 on Build GRCh38
rs114841105 Chr.10: 70146528-70146528 on Build GRCh38
rs568392077 Chr.10: 70146592-70146592 on Build GRCh38
rs539816274 Chr.10: 70146694-70146694 on Build GRCh38
rs764998660 Chr.10: 70146632-70146632 on Build GRCh38
rs763858383 Chr.10: 70146508-70146508 on Build GRCh38
rs548470278 Chr.10: 70146645-70146646 on Build GRCh38
rs774482910 Chr.10: 70146600-70146600 on Build GRCh38
rs535376729 Chr.10: 70146607-70146607 on Build GRCh38
rs566766159 Chr.10: 70146672-70146672 on Build GRCh38
rs867857641 Chr.10: 70146670-70146670 on Build GRCh38
rs530283518 Chr.10: 70146482-70146482 on Build GRCh38
rs762665059 Chr.10: 70146637-70146637 on Build GRCh38
rs755514668 Chr.10: 70146554-70146554 on Build GRCh38

View Full Product Details