Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57135736-57135979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZIM3
Entrez Gene ID
114026
Gene Name
zinc finger imprinted 3
Gene Aliases
ZNF657
UniGene
Hs.348622
Gene Chromosome Location
Chr.19: 57134096-57145202 on Build GRCh38
Transcripts
NM_052882.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7115530 c.547C>G p.Q183E Chr.19: 57135790-57135790 on Build GRCh38
3540342 c.553C>T p.H185Y Chr.19: 57135784-57135784 on Build GRCh38
393391 c.564G>A p.R188R Chr.19: 57135773-57135773 on Build GRCh38
6354553 c.399T>C p.D133D Chr.19: 57135938-57135938 on Build GRCh38
4552424 c.558G>T p.L186L Chr.19: 57135779-57135779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918355959 Chr.19: 57135903-57135903 on Build GRCh38
rs74974582 Chr.19: 57135843-57135843 on Build GRCh38
rs778337788 Chr.19: 57135873-57135873 on Build GRCh38
rs969989758 Chr.19: 57135803-57135803 on Build GRCh38
rs557421274 Chr.19: 57135796-57135796 on Build GRCh38
rs753624102 Chr.19: 57135888-57135888 on Build GRCh38
rs140629664 Chr.19: 57135807-57135807 on Build GRCh38
rs7252632 Chr.19: 57135868-57135868 on Build GRCh38
rs545311281 Chr.19: 57135917-57135917 on Build GRCh38
rs770606177 Chr.19: 57135918-57135918 on Build GRCh38
rs764954740 Chr.19: 57135770-57135770 on Build GRCh38
rs745523530 Chr.19: 57135945-57135945 on Build GRCh38
rs375178643 Chr.19: 57135898-57135898 on Build GRCh38
rs763573412 Chr.19: 57135865-57135865 on Build GRCh38
rs368721312 Chr.19: 57135786-57135786 on Build GRCh38
rs577588112 Chr.19: 57135841-57135841 on Build GRCh38
rs372146101 Chr.19: 57135800-57135800 on Build GRCh38
rs147553820 Chr.19: 57135861-57135861 on Build GRCh38
rs144475735 Chr.19: 57135849-57135849 on Build GRCh38
rs768566336 Chr.19: 57135785-57135785 on Build GRCh38
rs1039143026 Chr.19: 57135952-57135952 on Build GRCh38
rs777032999 Chr.19: 57135846-57135846 on Build GRCh38
rs140762229 Chr.19: 57135950-57135950 on Build GRCh38
rs147848892 Chr.19: 57135850-57135850 on Build GRCh38
rs779864336 Chr.19: 57135768-57135768 on Build GRCh38
rs753449854 Chr.19: 57135811-57135811 on Build GRCh38
rs760335482 Chr.19: 57135798-57135798 on Build GRCh38
rs758265716 Chr.19: 57135869-57135869 on Build GRCh38
rs776217114 Chr.19: 57135942-57135942 on Build GRCh38
rs752256791 Chr.19: 57135901-57135901 on Build GRCh38
rs78230615 Chr.19: 57135899-57135899 on Build GRCh38
rs918762029 Chr.19: 57135893-57135893 on Build GRCh38
rs371637635 Chr.19: 57135882-57135882 on Build GRCh38
rs368225214 Chr.19: 57135911-57135911 on Build GRCh38
rs754701418 Chr.19: 57135889-57135889 on Build GRCh38
rs1048580081 Chr.19: 57135935-57135935 on Build GRCh38
rs144011680 Chr.19: 57135795-57135795 on Build GRCh38
rs778229869 Chr.19: 57135874-57135888 on Build GRCh38
rs765961543 Chr.19: 57135936-57135936 on Build GRCh38

View Full Product Details