Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88681144-88681387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAI3
Entrez Gene ID
333929
Gene Name
snail family transcriptional repressor 3
Gene Aliases
SMUC, SNAIL3, ZNF293, Zfp293
UniGene
Hs.673548
Gene Chromosome Location
Chr.16: 88677682-88686474 on Build GRCh38
Transcripts
NM_178310.3 NR_024399.1

Gene Symbol
SNAI3-AS1
Entrez Gene ID
197187
Gene Name
SNAI3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.499548
Gene Chromosome Location
Chr.16: 88663373-88687186 on Build GRCh38
Transcripts
NM_178310.3 NR_024399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3889129 c.514C>T p.Q172* Chr.16: 88681277-88681277 on Build GRCh38
5811823 c.553T>A p.C185S Chr.16: 88681238-88681238 on Build GRCh38
4393720 c.617C>T p.T206M Chr.16: 88681174-88681174 on Build GRCh38
323496 c.541C>T p.R181C Chr.16: 88681250-88681250 on Build GRCh38
6785869 c.424G>T p.A142S Chr.16: 88681367-88681367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754015883 Chr.16: 88681215-88681215 on Build GRCh38
rs777273672 Chr.16: 88681357-88681357 on Build GRCh38
rs746750417 Chr.16: 88681229-88681229 on Build GRCh38
rs767802631 Chr.16: 88681245-88681245 on Build GRCh38
rs761184798 Chr.16: 88681331-88681331 on Build GRCh38
rs148592286 Chr.16: 88681232-88681232 on Build GRCh38
rs373207558 Chr.16: 88681365-88681365 on Build GRCh38
rs868446048 Chr.16: 88681337-88681337 on Build GRCh38
rs770478914 Chr.16: 88681312-88681312 on Build GRCh38
rs376634134 Chr.16: 88681283-88681283 on Build GRCh38
rs765898620 Chr.16: 88681243-88681244 on Build GRCh38
rs755124801 Chr.16: 88681269-88681269 on Build GRCh38
rs1022293111 Chr.16: 88681342-88681342 on Build GRCh38
rs894914992 Chr.16: 88681203-88681203 on Build GRCh38
rs868782571 Chr.16: 88681288-88681288 on Build GRCh38
rs766794910 Chr.16: 88681339-88681339 on Build GRCh38
rs888663892 Chr.16: 88681237-88681237 on Build GRCh38
rs532485625 Chr.16: 88681250-88681250 on Build GRCh38
rs780389593 Chr.16: 88681209-88681209 on Build GRCh38
rs758401834 Chr.16: 88681292-88681292 on Build GRCh38
rs750498928 Chr.16: 88681282-88681283 on Build GRCh38
rs765348985 Chr.16: 88681344-88681344 on Build GRCh38
rs150708380 Chr.16: 88681233-88681233 on Build GRCh38
rs754050466 Chr.16: 88681253-88681253 on Build GRCh38
rs758153179 Chr.16: 88681226-88681226 on Build GRCh38
rs577444696 Chr.16: 88681358-88681358 on Build GRCh38
rs750106792 Chr.16: 88681364-88681364 on Build GRCh38
rs757042660 Chr.16: 88681299-88681299 on Build GRCh38
rs146468110 Chr.16: 88681346-88681346 on Build GRCh38
rs780826279 Chr.16: 88681301-88681301 on Build GRCh38
rs372012683 Chr.16: 88681167-88681167 on Build GRCh38
rs536470345 Chr.16: 88681327-88681327 on Build GRCh38
rs771743116 Chr.16: 88681352-88681352 on Build GRCh38
rs142916652 Chr.16: 88681280-88681280 on Build GRCh38
rs775024169 Chr.16: 88681313-88681313 on Build GRCh38
rs557914224 Chr.16: 88681180-88681180 on Build GRCh38
rs137881385 Chr.16: 88681345-88681345 on Build GRCh38
rs755092009 Chr.16: 88681223-88681223 on Build GRCh38
rs369871750 Chr.16: 88681321-88681321 on Build GRCh38
rs770619254 Chr.16: 88681231-88681231 on Build GRCh38
rs142703619 Chr.16: 88681351-88681351 on Build GRCh38
rs748615467 Chr.16: 88681296-88681296 on Build GRCh38
rs372312758 Chr.16: 88681236-88681236 on Build GRCh38
rs773662551 Chr.16: 88681249-88681249 on Build GRCh38
rs1028338131 Chr.16: 88681318-88681318 on Build GRCh38
rs769316553 Chr.16: 88681310-88681310 on Build GRCh38
rs777550261 Chr.16: 88681295-88681295 on Build GRCh38
rs149676512 Chr.16: 88681184-88681184 on Build GRCh38
rs532228901 Chr.16: 88681360-88681360 on Build GRCh38
rs145862767 Chr.16: 88681227-88681227 on Build GRCh38
rs375859734 Chr.16: 88681311-88681311 on Build GRCh38
rs1048765656 Chr.16: 88681234-88681234 on Build GRCh38
rs368227192 Chr.16: 88681224-88681224 on Build GRCh38
rs766762396 Chr.16: 88681251-88681251 on Build GRCh38
rs147711305 Chr.16: 88681330-88681330 on Build GRCh38
rs759796565 Chr.16: 88681341-88681341 on Build GRCh38
rs202205064 Chr.16: 88681303-88681303 on Build GRCh38
rs142561823 Chr.16: 88681340-88681340 on Build GRCh38
rs989652707 Chr.16: 88681166-88681166 on Build GRCh38
rs752056919 Chr.16: 88681169-88681169 on Build GRCh38
rs762331912 Chr.16: 88681240-88681240 on Build GRCh38
rs139790709 Chr.16: 88681174-88681174 on Build GRCh38

View Full Product Details