Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81672580-81672831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC137
Entrez Gene ID
339230
Gene Name
coiled-coil domain containing 137
Gene Aliases
RaRF
UniGene
Hs.744125
Gene Chromosome Location
Chr.17: 81666697-81673907 on Build GRCh38
Transcripts
NM_199287.2 XM_011524738.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3821003 c.784T>C p.Y262H Chr.17: 81672618-81672618 on Build GRCh38
7359769 c.845G>A p.R282Q Chr.17: 81672679-81672679 on Build GRCh38
6426566 c.870A>G p.*290W Chr.17: 81672704-81672704 on Build GRCh38
4646571 c.839C>T p.T280I Chr.17: 81672673-81672673 on Build GRCh38
1610979 c.866T>C p.L289P Chr.17: 81672700-81672700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778907421 Chr.17: 81672613-81672613 on Build GRCh38
rs749604295 Chr.17: 81672753-81672753 on Build GRCh38
rs776318029 Chr.17: 81672714-81672714 on Build GRCh38
rs769706064 Chr.17: 81672712-81672713 on Build GRCh38
rs752481036 Chr.17: 81672657-81672657 on Build GRCh38
rs370516445 Chr.17: 81672679-81672679 on Build GRCh38
rs777883914 Chr.17: 81672623-81672623 on Build GRCh38
rs775314842 Chr.17: 81672717-81672717 on Build GRCh38
rs781395970 Chr.17: 81672626-81672626 on Build GRCh38
rs772733106 Chr.17: 81672644-81672644 on Build GRCh38
rs780376548 Chr.17: 81672689-81672689 on Build GRCh38
rs560097723 Chr.17: 81672800-81672800 on Build GRCh38
rs965539168 Chr.17: 81672640-81672640 on Build GRCh38
rs993276730 Chr.17: 81672705-81672705 on Build GRCh38
rs753648466 Chr.17: 81672604-81672604 on Build GRCh38
rs770557421 Chr.17: 81672701-81672701 on Build GRCh38
rs373340939 Chr.17: 81672647-81672647 on Build GRCh38
rs201048025 Chr.17: 81672726-81672726 on Build GRCh38
rs377260728 Chr.17: 81672653-81672653 on Build GRCh38
rs749640342 Chr.17: 81672690-81672690 on Build GRCh38
rs753561494 Chr.17: 81672649-81672649 on Build GRCh38
rs781308470 Chr.17: 81672677-81672677 on Build GRCh38
rs756280691 Chr.17: 81672732-81672732 on Build GRCh38
rs766065168 Chr.17: 81672648-81672648 on Build GRCh38
rs145262044 Chr.17: 81672731-81672731 on Build GRCh38
rs1012518483 Chr.17: 81672765-81672765 on Build GRCh38
rs534087953 Chr.17: 81672664-81672664 on Build GRCh38
rs769092974 Chr.17: 81672637-81672637 on Build GRCh38
rs375426067 Chr.17: 81672625-81672625 on Build GRCh38
rs373227595 Chr.17: 81672660-81672660 on Build GRCh38
rs1043822103 Chr.17: 81672719-81672719 on Build GRCh38
rs1036493982 Chr.17: 81672609-81672609 on Build GRCh38
rs757253184 Chr.17: 81672727-81672727 on Build GRCh38
rs752673813 Chr.17: 81672615-81672615 on Build GRCh38
rs754827508 Chr.17: 81672611-81672611 on Build GRCh38
rs777603327 Chr.17: 81672661-81672661 on Build GRCh38
rs747081294 Chr.17: 81672624-81672624 on Build GRCh38
rs61742298 Chr.17: 81672654-81672654 on Build GRCh38
rs1023818265 Chr.17: 81672651-81672651 on Build GRCh38
rs930834715 Chr.17: 81672759-81672759 on Build GRCh38
rs556023046 Chr.17: 81672695-81672695 on Build GRCh38
rs757342170 Chr.17: 81672666-81672666 on Build GRCh38
rs750563919 Chr.17: 81672631-81672632 on Build GRCh38
rs770130955 Chr.17: 81672629-81672629 on Build GRCh38
rs748146095 Chr.17: 81672637-81672638 on Build GRCh38
rs148449642 Chr.17: 81672694-81672694 on Build GRCh38
rs902548396 Chr.17: 81672608-81672608 on Build GRCh38
rs371255437 Chr.17: 81672716-81672716 on Build GRCh38
rs775901084 Chr.17: 81672630-81672630 on Build GRCh38
rs780174434 Chr.17: 81672750-81672750 on Build GRCh38
rs112699648 Chr.17: 81672636-81672636 on Build GRCh38
rs34921873 Chr.17: 81672601-81672602 on Build GRCh38
rs11546631 Chr.17: 81672678-81672678 on Build GRCh38
rs373358953 Chr.17: 81672691-81672691 on Build GRCh38
rs759237684 Chr.17: 81672715-81672715 on Build GRCh38
rs374379454 Chr.17: 81672730-81672730 on Build GRCh38

View Full Product Details