Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3766508-3766749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC27
Entrez Gene ID
148870
Gene Name
coiled-coil domain containing 27
Gene Aliases
-
UniGene
Hs.348700
Gene Chromosome Location
Chr.1: 3752398-3771645 on Build GRCh38
Transcripts
NM_152492.2 XM_011540780.1 XM_017000402.1 XM_017000403.1 XM_017000404.1 XM_017000405.1 XM_017000406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3489073 c.1466G>A p.R489Q Chr.1: 3766548-3766548 on Build GRCh38
3944015 c.1525C>T p.R509* Chr.1: 3766607-3766607 on Build GRCh38
7413293 c.1498C>A p.L500I Chr.1: 3766580-3766580 on Build GRCh38
6372301 c.1498C>T p.L500F Chr.1: 3766580-3766580 on Build GRCh38
3805079 c.1504G>A p.E502K Chr.1: 3766586-3766586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374880789 Chr.1: 3766629-3766629 on Build GRCh38
rs368645329 Chr.1: 3766702-3766702 on Build GRCh38
rs374173047 Chr.1: 3766640-3766640 on Build GRCh38
rs1028158521 Chr.1: 3766584-3766584 on Build GRCh38
rs1037426894 Chr.1: 3766533-3766533 on Build GRCh38
rs774182371 Chr.1: 3766662-3766662 on Build GRCh38
rs371836549 Chr.1: 3766627-3766627 on Build GRCh38
rs139337972 Chr.1: 3766573-3766573 on Build GRCh38
rs781477872 Chr.1: 3766626-3766626 on Build GRCh38
rs565221821 Chr.1: 3766691-3766691 on Build GRCh38
rs71634363 Chr.1: 3766712-3766712 on Build GRCh38
rs567850503 Chr.1: 3766730-3766730 on Build GRCh38
rs770270085 Chr.1: 3766630-3766630 on Build GRCh38
rs748800417 Chr.1: 3766645-3766645 on Build GRCh38
rs368925895 Chr.1: 3766652-3766652 on Build GRCh38
rs531124757 Chr.1: 3766722-3766722 on Build GRCh38
rs146467132 Chr.1: 3766609-3766609 on Build GRCh38
rs139864478 Chr.1: 3766729-3766729 on Build GRCh38
rs144089943 Chr.1: 3766595-3766595 on Build GRCh38
rs142677270 Chr.1: 3766547-3766547 on Build GRCh38
rs760999663 Chr.1: 3766678-3766678 on Build GRCh38
rs767384697 Chr.1: 3766548-3766548 on Build GRCh38
rs751737604 Chr.1: 3766615-3766615 on Build GRCh38
rs766330200 Chr.1: 3766582-3766582 on Build GRCh38
rs760475265 Chr.1: 3766575-3766575 on Build GRCh38
rs757622717 Chr.1: 3766624-3766624 on Build GRCh38
rs774125345 Chr.1: 3766536-3766536 on Build GRCh38
rs776258059 Chr.1: 3766575-3766575 on Build GRCh38
rs758447306 Chr.1: 3766608-3766608 on Build GRCh38

View Full Product Details