Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128895239-128895486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD9
Entrez Gene ID
28976
Gene Name
acyl-CoA dehydrogenase family member 9
Gene Aliases
NPD002
UniGene
Hs.567482
Gene Chromosome Location
Chr.3: 128879490-128916075 on Build GRCh38
Transcripts
NM_014049.4 NR_033426.1 XM_017006264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1038010 c.453+1G>A p.? Chr.3: 128895417-128895417 on Build GRCh38
5059275 c.352C>A p.L118M Chr.3: 128895315-128895315 on Build GRCh38
4490372 c.362C>T p.S121F Chr.3: 128895325-128895325 on Build GRCh38
3759716 c.379A>C p.R127R Chr.3: 128895342-128895342 on Build GRCh38
1418779 c.385G>A p.G129R Chr.3: 128895348-128895348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs863224844 Chr.3: 128895322-128895322 on Build GRCh38
rs764894010 Chr.3: 128895375-128895375 on Build GRCh38
rs766082543 Chr.3: 128895345-128895345 on Build GRCh38
rs1683791 Chr.3: 128895290-128895290 on Build GRCh38
rs768769460 Chr.3: 128895408-128895408 on Build GRCh38
rs779532712 Chr.3: 128895300-128895303 on Build GRCh38
rs879062561 Chr.3: 128895363-128895363 on Build GRCh38
rs199919500 Chr.3: 128895424-128895424 on Build GRCh38
rs753486926 Chr.3: 128895362-128895362 on Build GRCh38
rs774267812 Chr.3: 128895308-128895308 on Build GRCh38
rs202119704 Chr.3: 128895405-128895405 on Build GRCh38
rs758794781 Chr.3: 128895422-128895423 on Build GRCh38
rs368396605 Chr.3: 128895394-128895394 on Build GRCh38
rs775382898 Chr.3: 128895269-128895269 on Build GRCh38
rs565078746 Chr.3: 128895371-128895371 on Build GRCh38
rs780292473 Chr.3: 128895402-128895402 on Build GRCh38
rs1017729892 Chr.3: 128895270-128895270 on Build GRCh38
rs761798471 Chr.3: 128895321-128895321 on Build GRCh38
rs1680778 Chr.3: 128895342-128895342 on Build GRCh38
rs754503262 Chr.3: 128895370-128895370 on Build GRCh38
rs1007730431 Chr.3: 128895268-128895268 on Build GRCh38
rs878951917 Chr.3: 128895423-128895423 on Build GRCh38
rs760973317 Chr.3: 128895338-128895338 on Build GRCh38
rs757281259 Chr.3: 128895277-128895277 on Build GRCh38
rs973931566 Chr.3: 128895303-128895303 on Build GRCh38
rs539622467 Chr.3: 128895385-128895385 on Build GRCh38
rs368762565 Chr.3: 128895391-128895391 on Build GRCh38
rs767243207 Chr.3: 128895334-128895334 on Build GRCh38
rs749365099 Chr.3: 128895271-128895271 on Build GRCh38
rs752053336 Chr.3: 128895381-128895381 on Build GRCh38
rs370469605 Chr.3: 128895438-128895438 on Build GRCh38
rs781653464 Chr.3: 128895390-128895390 on Build GRCh38
rs140225132 Chr.3: 128895395-128895395 on Build GRCh38
rs774429198 Chr.3: 128895415-128895415 on Build GRCh38
rs772947137 Chr.3: 128895449-128895449 on Build GRCh38
rs1004471164 Chr.3: 128895467-128895467 on Build GRCh38

View Full Product Details