Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 58612667-58612932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP91
Entrez Gene ID
80829
Gene Name
ZFP91 zinc finger protein
Gene Aliases
DMS-8, DSM-8, FKSG11, PZF, ZFP-91, ZNF757
UniGene
Hs.524920
Gene Chromosome Location
Chr.11: 58579114-58621550 on Build GRCh38
Transcripts
NM_001197051.1 NM_053023.4 NR_024091.1

Gene Symbol
ZFP91-CNTF
Entrez Gene ID
386607
Gene Name
ZFP91-CNTF readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.524920
Gene Chromosome Location
Chr.11: 58579114-58625733 on Build GRCh38
Transcripts
NM_001197051.1 NM_053023.4 NR_024091.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2034715 c.935G>A p.R312H Chr.11: 58612788-58612788 on Build GRCh38
2034714 c.935G>A p.R312H Chr.11: 58612788-58612788 on Build GRCh38
1233422 c.977G>A p.R326H Chr.11: 58612830-58612830 on Build GRCh38
6826857 c.949G>A p.G317R Chr.11: 58612802-58612802 on Build GRCh38
1233423 c.977G>A p.R326H Chr.11: 58612830-58612830 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751398662 Chr.11: 58612871-58612871 on Build GRCh38
rs547987441 Chr.11: 58612720-58612721 on Build GRCh38
rs536090878 Chr.11: 58612720-58612720 on Build GRCh38
rs796611822 Chr.11: 58612720-58612721 on Build GRCh38
rs780797656 Chr.11: 58612886-58612886 on Build GRCh38
rs773698440 Chr.11: 58612712-58612713 on Build GRCh38
rs375153605 Chr.11: 58612897-58612897 on Build GRCh38
rs397730486 Chr.11: 58612731-58612732 on Build GRCh38
rs137938939 Chr.11: 58612700-58612700 on Build GRCh38
rs775928242 Chr.11: 58612733-58612733 on Build GRCh38
rs777042652 Chr.11: 58612798-58612798 on Build GRCh38
rs763893655 Chr.11: 58612731-58612737 on Build GRCh38
rs930733574 Chr.11: 58612776-58612776 on Build GRCh38
rs761952963 Chr.11: 58612801-58612801 on Build GRCh38
rs200535996 Chr.11: 58612732-58612732 on Build GRCh38
rs550620230 Chr.11: 58612709-58612709 on Build GRCh38
rs186579854 Chr.11: 58612890-58612890 on Build GRCh38
rs377342560 Chr.11: 58612856-58612856 on Build GRCh38
rs526217 Chr.11: 58612703-58612703 on Build GRCh38
rs2509923 Chr.11: 58612880-58612880 on Build GRCh38
rs1045541063 Chr.11: 58612755-58612755 on Build GRCh38
rs779519784 Chr.11: 58612813-58612813 on Build GRCh38
rs750064935 Chr.11: 58612733-58612734 on Build GRCh38
rs957534103 Chr.11: 58612895-58612895 on Build GRCh38
rs116276484 Chr.11: 58612903-58612903 on Build GRCh38
rs35224542 Chr.11: 58612719-58612720 on Build GRCh38
rs762867487 Chr.11: 58612730-58612737 on Build GRCh38
rs999122829 Chr.11: 58612860-58612860 on Build GRCh38
rs750312674 Chr.11: 58612852-58612852 on Build GRCh38
rs1012680697 Chr.11: 58612721-58612721 on Build GRCh38
rs751285295 Chr.11: 58612732-58612737 on Build GRCh38
rs779287453 Chr.11: 58612712-58612712 on Build GRCh38
rs769021922 Chr.11: 58612742-58612742 on Build GRCh38
rs756930201 Chr.11: 58612732-58612733 on Build GRCh38
rs370769781 Chr.11: 58612901-58612901 on Build GRCh38
rs373715398 Chr.11: 58612715-58612715 on Build GRCh38
rs894491023 Chr.11: 58612858-58612858 on Build GRCh38
rs754572593 Chr.11: 58612884-58612884 on Build GRCh38
rs201149041 Chr.11: 58612734-58612734 on Build GRCh38
rs766018663 Chr.11: 58612857-58612857 on Build GRCh38

View Full Product Details