Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113690714-113690951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS5
Entrez Gene ID
80975
Gene Name
transmembrane protease, serine 5
Gene Aliases
SPINESIN
UniGene
Hs.46720
Gene Chromosome Location
Chr.11: 113687546-113709811 on Build GRCh38
Transcripts
NM_001288749.1 NM_001288750.1 NM_001288751.1 NM_001288752.1 NM_030770.3 NR_110046.1 NR_110047.1 XM_017018366.1 XM_017018367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5837817 c.1043delG p.G348fs*62 Chr.11: 113690861-113690861 on Build GRCh38
923206 c.1038C>A p.G346G Chr.11: 113690866-113690866 on Build GRCh38
4734745 c.1037delG p.G346fs*64 Chr.11: 113690867-113690867 on Build GRCh38
7312753 c.989C>T p.P330L Chr.11: 113690915-113690915 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777598000 Chr.11: 113690914-113690914 on Build GRCh38
rs766611121 Chr.11: 113690923-113690929 on Build GRCh38
rs868666039 Chr.11: 113690861-113690861 on Build GRCh38
rs377473163 Chr.11: 113690817-113690817 on Build GRCh38
rs748335373 Chr.11: 113690904-113690904 on Build GRCh38
rs757272360 Chr.11: 113690809-113690809 on Build GRCh38
rs374278839 Chr.11: 113690915-113690915 on Build GRCh38
rs375585424 Chr.11: 113690872-113690872 on Build GRCh38
rs773312031 Chr.11: 113690881-113690881 on Build GRCh38
rs912588742 Chr.11: 113690795-113690795 on Build GRCh38
rs1044390829 Chr.11: 113690885-113690885 on Build GRCh38
rs202148348 Chr.11: 113690849-113690849 on Build GRCh38
rs753251919 Chr.11: 113690807-113690807 on Build GRCh38
rs748199416 Chr.11: 113690857-113690857 on Build GRCh38
rs375638868 Chr.11: 113690901-113690901 on Build GRCh38
rs773608991 Chr.11: 113690767-113690767 on Build GRCh38
rs746245364 Chr.11: 113690929-113690929 on Build GRCh38
rs568115704 Chr.11: 113690882-113690882 on Build GRCh38
rs367586238 Chr.11: 113690883-113690883 on Build GRCh38
rs117055692 Chr.11: 113690928-113690928 on Build GRCh38
rs761160755 Chr.11: 113690806-113690806 on Build GRCh38
rs773712183 Chr.11: 113690868-113690868 on Build GRCh38
rs186244114 Chr.11: 113690887-113690887 on Build GRCh38
rs539374810 Chr.11: 113690890-113690890 on Build GRCh38
rs1057371489 Chr.11: 113690867-113690867 on Build GRCh38
rs368166557 Chr.11: 113690893-113690893 on Build GRCh38
rs201233178 Chr.11: 113690894-113690894 on Build GRCh38
rs763074958 Chr.11: 113690883-113690884 on Build GRCh38
rs1056900057 Chr.11: 113690803-113690803 on Build GRCh38
rs368573232 Chr.11: 113690913-113690913 on Build GRCh38
rs189987760 Chr.11: 113690863-113690863 on Build GRCh38
rs771393853 Chr.11: 113690917-113690917 on Build GRCh38
rs45570834 Chr.11: 113690760-113690760 on Build GRCh38
rs535175585 Chr.11: 113690916-113690916 on Build GRCh38
rs546675449 Chr.11: 113690796-113690796 on Build GRCh38
rs560564391 Chr.11: 113690805-113690805 on Build GRCh38
rs397897481 Chr.11: 113690763-113690763 on Build GRCh38
rs1053987915 Chr.11: 113690853-113690853 on Build GRCh38
rs180841444 Chr.11: 113690884-113690884 on Build GRCh38
rs371177905 Chr.11: 113690822-113690822 on Build GRCh38
rs924397124 Chr.11: 113690830-113690830 on Build GRCh38
rs779939238 Chr.11: 113690827-113690827 on Build GRCh38
rs781373229 Chr.11: 113690852-113690852 on Build GRCh38
rs778396904 Chr.11: 113690818-113690818 on Build GRCh38
rs771340675 Chr.11: 113690874-113690874 on Build GRCh38
rs1042770456 Chr.11: 113690744-113690744 on Build GRCh38

View Full Product Details