Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2042592-2042839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MKNK2
Entrez Gene ID
2872
Gene Name
MAP kinase interacting serine/threonine kinase 2
Gene Aliases
GPRK7, MNK2
UniGene
Hs.515032
Gene Chromosome Location
Chr.19: 2037471-2051244 on Build GRCh38
Transcripts
NM_017572.3 NM_199054.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3701441 c.609C>A p.H203Q Chr.19: 2042652-2042652 on Build GRCh38
4154115 c.563A>G p.D188G Chr.19: 2042801-2042801 on Build GRCh38
4154114 c.563A>G p.D188G Chr.19: 2042801-2042801 on Build GRCh38
4995860 c.562G>T p.D188Y Chr.19: 2042802-2042802 on Build GRCh38
7210565 c.613G>T p.D205Y Chr.19: 2042648-2042648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757080449 Chr.19: 2042800-2042800 on Build GRCh38
rs201595763 Chr.19: 2042704-2042704 on Build GRCh38
rs758257789 Chr.19: 2042690-2042692 on Build GRCh38
rs753681155 Chr.19: 2042751-2042751 on Build GRCh38
rs369980643 Chr.19: 2042623-2042623 on Build GRCh38
rs546219897 Chr.19: 2042640-2042640 on Build GRCh38
rs377396725 Chr.19: 2042722-2042722 on Build GRCh38
rs757298708 Chr.19: 2042778-2042778 on Build GRCh38
rs1039116356 Chr.19: 2042720-2042720 on Build GRCh38
rs866566834 Chr.19: 2042796-2042796 on Build GRCh38
rs114543515 Chr.19: 2042736-2042736 on Build GRCh38
rs777710525 Chr.19: 2042752-2042753 on Build GRCh38
rs373373714 Chr.19: 2042687-2042687 on Build GRCh38
rs770978561 Chr.19: 2042686-2042686 on Build GRCh38
rs762377486 Chr.19: 2042732-2042732 on Build GRCh38
rs963639360 Chr.19: 2042667-2042667 on Build GRCh38
rs751119935 Chr.19: 2042739-2042739 on Build GRCh38
rs866513976 Chr.19: 2042645-2042645 on Build GRCh38
rs755883725 Chr.19: 2042638-2042638 on Build GRCh38
rs200707721 Chr.19: 2042745-2042745 on Build GRCh38
rs577423373 Chr.19: 2042815-2042815 on Build GRCh38
rs780126999 Chr.19: 2042799-2042799 on Build GRCh38
rs772072993 Chr.19: 2042791-2042791 on Build GRCh38
rs946143882 Chr.19: 2042698-2042698 on Build GRCh38
rs112345263 Chr.19: 2042665-2042665 on Build GRCh38
rs749249240 Chr.19: 2042671-2042671 on Build GRCh38
rs868825213 Chr.19: 2042650-2042650 on Build GRCh38
rs747255365 Chr.19: 2042811-2042811 on Build GRCh38
rs78002226 Chr.19: 2042664-2042664 on Build GRCh38
rs112650981 Chr.19: 2042764-2042764 on Build GRCh38
rs540485640 Chr.19: 2042816-2042816 on Build GRCh38
rs747787366 Chr.19: 2042646-2042646 on Build GRCh38
rs760908613 Chr.19: 2042707-2042707 on Build GRCh38
rs929462907 Chr.19: 2042658-2042658 on Build GRCh38
rs1016560874 Chr.19: 2042625-2042625 on Build GRCh38
rs778968779 Chr.19: 2042781-2042781 on Build GRCh38
rs1029503836 Chr.19: 2042678-2042678 on Build GRCh38
rs373634147 Chr.19: 2042812-2042812 on Build GRCh38
rs777484977 Chr.19: 2042750-2042750 on Build GRCh38
rs776969829 Chr.19: 2042729-2042729 on Build GRCh38
rs777556978 Chr.19: 2042669-2042669 on Build GRCh38
rs753105970 Chr.19: 2042782-2042782 on Build GRCh38
rs1032886639 Chr.19: 2042737-2042737 on Build GRCh38
rs72981432 Chr.19: 2042746-2042746 on Build GRCh38
rs745746481 Chr.19: 2042783-2042783 on Build GRCh38
rs772382243 Chr.19: 2042689-2042689 on Build GRCh38
rs75160224 Chr.19: 2042747-2042747 on Build GRCh38

View Full Product Details