Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109750242-109750515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS8L3
Entrez Gene ID
79574
Gene Name
EPS8 like 3
Gene Aliases
EPS8R3
UniGene
Hs.485352
Gene Chromosome Location
Chr.1: 109750080-109764022 on Build GRCh38
Transcripts
NM_001319952.1 NM_024526.3 NM_133181.3 NM_139053.2 XM_011542132.1 XM_011542133.1 XM_011542134.2 XM_011542135.2 XM_017002328.1 XM_017002329.1

SNP Detail

SNP ID Chromosome Location
rs1037847772 Chr.1: 109750295-109750295 on Build GRCh38
rs191322577 Chr.1: 109750323-109750323 on Build GRCh38
rs928945568 Chr.1: 109750309-109750309 on Build GRCh38
rs148530481 Chr.1: 109750317-109750317 on Build GRCh38
rs779512283 Chr.1: 109750361-109750361 on Build GRCh38
rs370759963 Chr.1: 109750403-109750403 on Build GRCh38
rs201481368 Chr.1: 109750357-109750357 on Build GRCh38
rs956358977 Chr.1: 109750317-109750317 on Build GRCh38
rs1016482668 Chr.1: 109750366-109750366 on Build GRCh38
rs771170860 Chr.1: 109750397-109750397 on Build GRCh38
rs376835702 Chr.1: 109750424-109750424 on Build GRCh38
rs771352122 Chr.1: 109750398-109750398 on Build GRCh38
rs974108949 Chr.1: 109750479-109750479 on Build GRCh38
rs1029635483 Chr.1: 109750458-109750458 on Build GRCh38
rs769590116 Chr.1: 109750414-109750414 on Build GRCh38
rs752714187 Chr.1: 109750375-109750375 on Build GRCh38
rs997466496 Chr.1: 109750331-109750331 on Build GRCh38
rs201801498 Chr.1: 109750356-109750356 on Build GRCh38
rs747164126 Chr.1: 109750386-109750386 on Build GRCh38
rs1030254935 Chr.1: 109750337-109750337 on Build GRCh38
rs745510589 Chr.1: 109750339-109750339 on Build GRCh38
rs954296116 Chr.1: 109750462-109750462 on Build GRCh38
rs1012760851 Chr.1: 109750344-109750344 on Build GRCh38
rs566805633 Chr.1: 109750438-109750438 on Build GRCh38
rs375211281 Chr.1: 109750406-109750406 on Build GRCh38
rs775568104 Chr.1: 109750418-109750418 on Build GRCh38
rs766784623 Chr.1: 109750348-109750348 on Build GRCh38
rs371180538 Chr.1: 109750430-109750430 on Build GRCh38
rs77091584 Chr.1: 109750396-109750396 on Build GRCh38
rs75119925 Chr.1: 109750302-109750302 on Build GRCh38
rs998582998 Chr.1: 109750436-109750436 on Build GRCh38
rs561101553 Chr.1: 109750351-109750351 on Build GRCh38
rs879231527 Chr.1: 109750329-109750329 on Build GRCh38

View Full Product Details