Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 2040438-2040705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPH1
Entrez Gene ID
1801
Gene Name
diphthamide biosynthesis 1
Gene Aliases
DEDSSH, DPH2L, DPH2L1, OVCA1
UniGene
Hs.513856
Gene Chromosome Location
Chr.17: 2030137-2043431 on Build GRCh38
Transcripts
NM_001383.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7103352 c.932C>T p.S311F Chr.17: 2040515-2040515 on Build GRCh38
143518 c.949_950GG>AC p.G317T Chr.17: 2040532-2040533 on Build GRCh38
5924072 c.1012G>A p.E338K Chr.17: 2040595-2040595 on Build GRCh38
3421331 c.935G>A p.R312Q Chr.17: 2040518-2040518 on Build GRCh38
6229745 c.980A>G p.E327G Chr.17: 2040563-2040563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778114854 Chr.17: 2040644-2040644 on Build GRCh38
rs748598394 Chr.17: 2040543-2040543 on Build GRCh38
rs764032731 Chr.17: 2040504-2040504 on Build GRCh38
rs35394823 Chr.17: 2040586-2040586 on Build GRCh38
rs755153813 Chr.17: 2040629-2040629 on Build GRCh38
rs760608696 Chr.17: 2040501-2040501 on Build GRCh38
rs766652021 Chr.17: 2040604-2040604 on Build GRCh38
rs774119740 Chr.17: 2040478-2040478 on Build GRCh38
rs753929581 Chr.17: 2040476-2040477 on Build GRCh38
rs766395606 Chr.17: 2040672-2040672 on Build GRCh38
rs745550380 Chr.17: 2040548-2040548 on Build GRCh38
rs1049924857 Chr.17: 2040596-2040596 on Build GRCh38
rs182574496 Chr.17: 2040483-2040483 on Build GRCh38
rs867798707 Chr.17: 2040555-2040555 on Build GRCh38
rs760232816 Chr.17: 2040570-2040570 on Build GRCh38
rs777703572 Chr.17: 2040477-2040477 on Build GRCh38
rs1018546327 Chr.17: 2040666-2040666 on Build GRCh38
rs985671603 Chr.17: 2040663-2040663 on Build GRCh38
rs944023226 Chr.17: 2040533-2040533 on Build GRCh38
rs1035355788 Chr.17: 2040637-2040637 on Build GRCh38
rs767034803 Chr.17: 2040485-2040485 on Build GRCh38
rs750726709 Chr.17: 2040523-2040523 on Build GRCh38
rs200849211 Chr.17: 2040562-2040562 on Build GRCh38
rs777949712 Chr.17: 2040547-2040547 on Build GRCh38
rs753270530 Chr.17: 2040638-2040638 on Build GRCh38
rs1027127820 Chr.17: 2040659-2040659 on Build GRCh38
rs748488781 Chr.17: 2040460-2040460 on Build GRCh38
rs199664244 Chr.17: 2040544-2040544 on Build GRCh38
rs757405584 Chr.17: 2040512-2040525 on Build GRCh38
rs761637932 Chr.17: 2040592-2040592 on Build GRCh38
rs745879918 Chr.17: 2040521-2040521 on Build GRCh38
rs771643497 Chr.17: 2040553-2040553 on Build GRCh38
rs905500811 Chr.17: 2040618-2040618 on Build GRCh38
rs990620472 Chr.17: 2040529-2040529 on Build GRCh38
rs756537646 Chr.17: 2040643-2040643 on Build GRCh38
rs935437729 Chr.17: 2040576-2040576 on Build GRCh38
rs953012598 Chr.17: 2040661-2040661 on Build GRCh38
rs201891404 Chr.17: 2040531-2040531 on Build GRCh38
rs746136888 Chr.17: 2040556-2040559 on Build GRCh38
rs1043732960 Chr.17: 2040667-2040667 on Build GRCh38
rs370213255 Chr.17: 2040633-2040633 on Build GRCh38
rs2236375 Chr.17: 2040594-2040594 on Build GRCh38
rs61757361 Chr.17: 2040524-2040524 on Build GRCh38
rs749595519 Chr.17: 2040651-2040651 on Build GRCh38
rs768044127 Chr.17: 2040585-2040585 on Build GRCh38
rs371066962 Chr.17: 2040517-2040517 on Build GRCh38
rs961451063 Chr.17: 2040475-2040475 on Build GRCh38
rs994354309 Chr.17: 2040647-2040647 on Build GRCh38
rs751819245 Chr.17: 2040615-2040615 on Build GRCh38
rs781752622 Chr.17: 2040539-2040539 on Build GRCh38
rs565199226 Chr.17: 2040598-2040598 on Build GRCh38
rs764906514 Chr.17: 2040518-2040518 on Build GRCh38
rs985724114 Chr.17: 2040530-2040530 on Build GRCh38
rs779128948 Chr.17: 2040526-2040532 on Build GRCh38
rs755455275 Chr.17: 2040532-2040532 on Build GRCh38
rs757139451 Chr.17: 2040508-2040508 on Build GRCh38
rs770042759 Chr.17: 2040463-2040463 on Build GRCh38
rs375530492 Chr.17: 2040595-2040595 on Build GRCh38
rs774939632 Chr.17: 2040492-2040492 on Build GRCh38

View Full Product Details