Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 246634378-246634649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNST
Entrez Gene ID
163882
Gene Name
consortin, connexin sorting protein
Gene Aliases
C1orf71, PPP1R64
UniGene
Hs.368353
Gene Chromosome Location
Chr.1: 246566337-246668587 on Build GRCh38
Transcripts
NM_001139459.1 NM_152609.2 XM_005273081.3 XM_005273083.4 XM_011544110.2 XM_011544111.1 XM_011544112.1 XM_011544113.1 XM_011544114.2 XM_017000487.1 XM_017000488.1 XM_017000489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5846080 c.787C>T p.R263W Chr.1: 246634556-246634556 on Build GRCh38
4030467 c.771T>C p.N257N Chr.1: 246634540-246634540 on Build GRCh38
906852 c.709A>C p.K237Q Chr.1: 246634478-246634478 on Build GRCh38
906853 c.818+1_818+2insT p.? Chr.1: 246634588-246634589 on Build GRCh38
3486498 c.769A>G p.N257D Chr.1: 246634538-246634538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765380022 Chr.1: 246634410-246634412 on Build GRCh38
rs780010747 Chr.1: 246634481-246634481 on Build GRCh38
rs901199178 Chr.1: 246634531-246634531 on Build GRCh38
rs376138645 Chr.1: 246634584-246634584 on Build GRCh38
rs367745311 Chr.1: 246634509-246634509 on Build GRCh38
rs373604546 Chr.1: 246634500-246634500 on Build GRCh38
rs552323655 Chr.1: 246634455-246634455 on Build GRCh38
rs777578194 Chr.1: 246634507-246634508 on Build GRCh38
rs752140039 Chr.1: 246634610-246634610 on Build GRCh38
rs548251611 Chr.1: 246634588-246634588 on Build GRCh38
rs201669819 Chr.1: 246634412-246634412 on Build GRCh38
rs768090992 Chr.1: 246634529-246634529 on Build GRCh38
rs763814011 Chr.1: 246634453-246634456 on Build GRCh38
rs532075825 Chr.1: 246634413-246634413 on Build GRCh38
rs11396823 Chr.1: 246634412-246634413 on Build GRCh38
rs746879740 Chr.1: 246634489-246634489 on Build GRCh38
rs775409438 Chr.1: 246634556-246634556 on Build GRCh38
rs752851657 Chr.1: 246634431-246634431 on Build GRCh38
rs935397683 Chr.1: 246634456-246634457 on Build GRCh38
rs756241220 Chr.1: 246634448-246634448 on Build GRCh38
rs371542664 Chr.1: 246634457-246634457 on Build GRCh38
rs761519634 Chr.1: 246634557-246634557 on Build GRCh38
rs767428428 Chr.1: 246634429-246634429 on Build GRCh38
rs147921633 Chr.1: 246634508-246634508 on Build GRCh38
rs776822854 Chr.1: 246634540-246634542 on Build GRCh38
rs760098376 Chr.1: 246634426-246634426 on Build GRCh38
rs993890203 Chr.1: 246634594-246634594 on Build GRCh38
rs1049720447 Chr.1: 246634601-246634601 on Build GRCh38
rs766239884 Chr.1: 246634577-246634577 on Build GRCh38
rs569002004 Chr.1: 246634514-246634514 on Build GRCh38
rs35650980 Chr.1: 246634408-246634408 on Build GRCh38
rs141101872 Chr.1: 246634564-246634564 on Build GRCh38
rs201963187 Chr.1: 246634411-246634412 on Build GRCh38
rs750150014 Chr.1: 246634585-246634585 on Build GRCh38
rs750239938 Chr.1: 246634456-246634456 on Build GRCh38
rs903952032 Chr.1: 246634437-246634437 on Build GRCh38
rs147112975 Chr.1: 246634521-246634521 on Build GRCh38
rs1045651679 Chr.1: 246634504-246634504 on Build GRCh38
rs890041764 Chr.1: 246634605-246634605 on Build GRCh38
rs749636732 Chr.1: 246634510-246634510 on Build GRCh38
rs372864418 Chr.1: 246634418-246634419 on Build GRCh38
rs754058519 Chr.1: 246634563-246634563 on Build GRCh38
rs896531661 Chr.1: 246634487-246634487 on Build GRCh38
rs200406172 Chr.1: 246634411-246634411 on Build GRCh38
rs754977780 Chr.1: 246634490-246634490 on Build GRCh38
rs781233322 Chr.1: 246634606-246634606 on Build GRCh38
rs371579273 Chr.1: 246634526-246634526 on Build GRCh38
rs764399186 Chr.1: 246634558-246634558 on Build GRCh38

View Full Product Details