Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90087029-90087296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDH2
Entrez Gene ID
3418
Gene Name
isocitrate dehydrogenase (NADP(+)) 2, mitochondrial
Gene Aliases
D2HGA2, ICD-M, IDH, IDHM, IDP, IDPM, mNADP-IDH
UniGene
Hs.596461
Gene Chromosome Location
Chr.15: 90083978-90102554 on Build GRCh38
Transcripts
NM_001289910.1 NM_001290114.1 NM_002168.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7403822 c.865_865delC p.L289fs*41 Chr.15: 90087214-90087214 on Build GRCh38
1375397 c.829G>A p.D277N Chr.15: 90087250-90087250 on Build GRCh38
6910840 c.851G>A p.W284* Chr.15: 90087228-90087228 on Build GRCh38
6278517 c.835G>A p.D279N Chr.15: 90087244-90087244 on Build GRCh38
6555222 c.927G>A p.K309K Chr.15: 90087152-90087152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778404157 Chr.15: 90087054-90087054 on Build GRCh38
rs773753491 Chr.15: 90087231-90087231 on Build GRCh38
rs113497619 Chr.15: 90087071-90087071 on Build GRCh38
rs938229610 Chr.15: 90087245-90087245 on Build GRCh38
rs751453863 Chr.15: 90087160-90087160 on Build GRCh38
rs763944027 Chr.15: 90087143-90087143 on Build GRCh38
rs765310294 Chr.15: 90087251-90087251 on Build GRCh38
rs142721369 Chr.15: 90087137-90087137 on Build GRCh38
rs371460407 Chr.15: 90087176-90087176 on Build GRCh38
rs779582884 Chr.15: 90087075-90087075 on Build GRCh38
rs767324925 Chr.15: 90087094-90087094 on Build GRCh38
rs774183637 Chr.15: 90087258-90087258 on Build GRCh38
rs138800371 Chr.15: 90087166-90087166 on Build GRCh38
rs538460237 Chr.15: 90087189-90087189 on Build GRCh38
rs761847749 Chr.15: 90087260-90087260 on Build GRCh38
rs772355218 Chr.15: 90087228-90087228 on Build GRCh38
rs754447918 Chr.15: 90087225-90087225 on Build GRCh38
rs773589925 Chr.15: 90087097-90087097 on Build GRCh38
rs752451868 Chr.15: 90087215-90087215 on Build GRCh38
rs759133024 Chr.15: 90087080-90087080 on Build GRCh38
rs772421080 Chr.15: 90087094-90087094 on Build GRCh38
rs748779376 Chr.15: 90087083-90087083 on Build GRCh38
rs946299599 Chr.15: 90087053-90087053 on Build GRCh38
rs202227794 Chr.15: 90087233-90087233 on Build GRCh38
rs762085634 Chr.15: 90087086-90087086 on Build GRCh38
rs149387924 Chr.15: 90087217-90087217 on Build GRCh38
rs749774631 Chr.15: 90087061-90087061 on Build GRCh38
rs767416694 Chr.15: 90087273-90087273 on Build GRCh38
rs16943901 Chr.15: 90087140-90087140 on Build GRCh38
rs776925988 Chr.15: 90087241-90087241 on Build GRCh38
rs766676533 Chr.15: 90087133-90087133 on Build GRCh38
rs748642079 Chr.15: 90087227-90087227 on Build GRCh38
rs779303729 Chr.15: 90087226-90087226 on Build GRCh38
rs780922457 Chr.15: 90087168-90087168 on Build GRCh38
rs759547195 Chr.15: 90087248-90087248 on Build GRCh38
rs951275836 Chr.15: 90087244-90087244 on Build GRCh38
rs771346231 Chr.15: 90087235-90087235 on Build GRCh38
rs760938795 Chr.15: 90087102-90087102 on Build GRCh38
rs769097040 Chr.15: 90087084-90087084 on Build GRCh38

View Full Product Details