Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48970039-48970304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCND1
Entrez Gene ID
3750
Gene Name
potassium voltage-gated channel subfamily D member 1
Gene Aliases
KV4.1
UniGene
Hs.55276
Gene Chromosome Location
Chr.X: 48960983-48972099 on Build GRCh38
Transcripts
NM_004979.5 XM_011543910.2 XM_017029508.1 XM_017029509.1 XM_017029510.1

SNP Detail

SNP ID Chromosome Location
rs782742345 Chr.X: 48970201-48970201 on Build GRCh38
rs782156359 Chr.X: 48970102-48970102 on Build GRCh38
rs781928762 Chr.X: 48970130-48970130 on Build GRCh38
rs782207073 Chr.X: 48970065-48970065 on Build GRCh38
rs181033799 Chr.X: 48970186-48970186 on Build GRCh38
rs368584539 Chr.X: 48970131-48970131 on Build GRCh38
rs966834990 Chr.X: 48970068-48970068 on Build GRCh38
rs782333585 Chr.X: 48970167-48970167 on Build GRCh38
rs782567521 Chr.X: 48970081-48970081 on Build GRCh38
rs782010774 Chr.X: 48970169-48970169 on Build GRCh38
rs782455528 Chr.X: 48970235-48970235 on Build GRCh38
rs782434162 Chr.X: 48970100-48970100 on Build GRCh38
rs200530989 Chr.X: 48970267-48970267 on Build GRCh38
rs782080524 Chr.X: 48970195-48970195 on Build GRCh38
rs781995145 Chr.X: 48970188-48970188 on Build GRCh38
rs782802983 Chr.X: 48970251-48970251 on Build GRCh38
rs781856558 Chr.X: 48970252-48970252 on Build GRCh38
rs782349071 Chr.X: 48970125-48970125 on Build GRCh38
rs782442631 Chr.X: 48970279-48970279 on Build GRCh38
rs909881285 Chr.X: 48970180-48970180 on Build GRCh38
rs782488264 Chr.X: 48970066-48970066 on Build GRCh38
rs1037547421 Chr.X: 48970229-48970229 on Build GRCh38
rs782104137 Chr.X: 48970194-48970194 on Build GRCh38
rs1050222371 Chr.X: 48970143-48970143 on Build GRCh38
rs782739049 Chr.X: 48970266-48970266 on Build GRCh38
rs782146641 Chr.X: 48970234-48970234 on Build GRCh38
rs782307815 Chr.X: 48970124-48970124 on Build GRCh38
rs782154287 Chr.X: 48970185-48970185 on Build GRCh38
rs782275593 Chr.X: 48970089-48970089 on Build GRCh38
rs782228279 Chr.X: 48970273-48970273 on Build GRCh38
rs782188441 Chr.X: 48970137-48970137 on Build GRCh38
rs976846418 Chr.X: 48970074-48970074 on Build GRCh38
rs782515541 Chr.X: 48970261-48970261 on Build GRCh38
rs368004864 Chr.X: 48970179-48970179 on Build GRCh38

View Full Product Details