Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41504679-41504952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LTK
Entrez Gene ID
4058
Gene Name
leukocyte receptor tyrosine kinase
Gene Aliases
TYK1
UniGene
Hs.434481
Gene Chromosome Location
Chr.15: 41503639-41513887 on Build GRCh38
Transcripts
NM_001135685.1 NM_002344.5 NM_206961.3 XM_011521556.2 XM_011521557.2 XM_017022181.1 XM_017022182.1 XM_017022183.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3886937 c.1922C>T p.S641F Chr.15: 41504788-41504788 on Build GRCh38
1559446 c.1850G>C p.R617P Chr.15: 41504860-41504860 on Build GRCh38
1662650 c.2032C>T p.R678C Chr.15: 41504861-41504861 on Build GRCh38
699877 c.2093G>A p.G698D Chr.15: 41504800-41504800 on Build GRCh38
961429 c.1930G>A p.D644N Chr.15: 41504780-41504780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749229003 Chr.15: 41504878-41504878 on Build GRCh38
rs748131904 Chr.15: 41504849-41504849 on Build GRCh38
rs780609118 Chr.15: 41504891-41504892 on Build GRCh38
rs747529266 Chr.15: 41504921-41504922 on Build GRCh38
rs170296 Chr.15: 41504703-41504703 on Build GRCh38
rs773974334 Chr.15: 41504902-41504902 on Build GRCh38
rs777699590 Chr.15: 41504738-41504738 on Build GRCh38
rs943837931 Chr.15: 41504776-41504776 on Build GRCh38
rs141045322 Chr.15: 41504848-41504848 on Build GRCh38
rs925548724 Chr.15: 41504734-41504734 on Build GRCh38
rs753466749 Chr.15: 41504739-41504743 on Build GRCh38
rs775228501 Chr.15: 41504914-41504914 on Build GRCh38
rs34619111 Chr.15: 41504930-41504930 on Build GRCh38
rs768340885 Chr.15: 41504769-41504769 on Build GRCh38
rs372800939 Chr.15: 41504782-41504782 on Build GRCh38
rs369369523 Chr.15: 41504763-41504764 on Build GRCh38
rs747851535 Chr.15: 41504861-41504861 on Build GRCh38
rs760889402 Chr.15: 41504916-41504916 on Build GRCh38
rs776270325 Chr.15: 41504820-41504820 on Build GRCh38
rs778383630 Chr.15: 41504794-41504794 on Build GRCh38
rs568286327 Chr.15: 41504715-41504715 on Build GRCh38
rs757739660 Chr.15: 41504924-41504924 on Build GRCh38
rs372223410 Chr.15: 41504868-41504868 on Build GRCh38
rs2278658 Chr.15: 41504773-41504773 on Build GRCh38
rs142798669 Chr.15: 41504860-41504860 on Build GRCh38
rs762498945 Chr.15: 41504846-41504846 on Build GRCh38
rs765647690 Chr.15: 41504752-41504753 on Build GRCh38
rs769581905 Chr.15: 41504840-41504840 on Build GRCh38
rs776555707 Chr.15: 41504770-41504770 on Build GRCh38
rs769949765 Chr.15: 41504851-41504851 on Build GRCh38
rs969592963 Chr.15: 41504708-41504708 on Build GRCh38
rs767036081 Chr.15: 41504911-41504911 on Build GRCh38
rs765269626 Chr.15: 41504774-41504774 on Build GRCh38
rs376599996 Chr.15: 41504880-41504880 on Build GRCh38
rs759123183 Chr.15: 41504910-41504910 on Build GRCh38
rs756553832 Chr.15: 41504791-41504791 on Build GRCh38
rs142986350 Chr.15: 41504781-41504781 on Build GRCh38
rs760412503 Chr.15: 41504764-41504764 on Build GRCh38
rs373383923 Chr.15: 41504921-41504921 on Build GRCh38
rs757829209 Chr.15: 41504855-41504855 on Build GRCh38
rs146757784 Chr.15: 41504856-41504856 on Build GRCh38
rs371910880 Chr.15: 41504744-41504754 on Build GRCh38
rs781684523 Chr.15: 41504733-41504733 on Build GRCh38
rs750815174 Chr.15: 41504757-41504758 on Build GRCh38
rs1049967337 Chr.15: 41504912-41504912 on Build GRCh38
rs751513576 Chr.15: 41504728-41504729 on Build GRCh38
rs912299788 Chr.15: 41504762-41504762 on Build GRCh38
rs766085406 Chr.15: 41504780-41504780 on Build GRCh38
rs748604191 Chr.15: 41504736-41504736 on Build GRCh38
rs535222095 Chr.15: 41504930-41504930 on Build GRCh38
rs369559975 Chr.15: 41504897-41504897 on Build GRCh38
rs201729483 Chr.15: 41504743-41504744 on Build GRCh38
rs746087075 Chr.15: 41504816-41504816 on Build GRCh38
rs749925163 Chr.15: 41504778-41504778 on Build GRCh38
rs771020055 Chr.15: 41504800-41504800 on Build GRCh38
rs781402616 Chr.15: 41504785-41504785 on Build GRCh38
rs377484254 Chr.15: 41504922-41504922 on Build GRCh38
rs758764755 Chr.15: 41504851-41504852 on Build GRCh38
rs762753546 Chr.15: 41504727-41504728 on Build GRCh38
rs764457664 Chr.15: 41504853-41504853 on Build GRCh38
rs575625623 Chr.15: 41504863-41504863 on Build GRCh38
rs140642102 Chr.15: 41504838-41504838 on Build GRCh38
rs773307999 Chr.15: 41504844-41504844 on Build GRCh38
rs766340702 Chr.15: 41504728-41504748 on Build GRCh38
rs562881917 Chr.15: 41504744-41504744 on Build GRCh38
rs753160475 Chr.15: 41504789-41504789 on Build GRCh38
rs1032660029 Chr.15: 41504798-41504798 on Build GRCh38
rs367709220 Chr.15: 41504737-41504737 on Build GRCh38
rs754355804 Chr.15: 41504854-41504854 on Build GRCh38
rs375465862 Chr.15: 41504799-41504799 on Build GRCh38
rs771109740 Chr.15: 41504755-41504755 on Build GRCh38
rs749415823 Chr.15: 41504754-41504754 on Build GRCh38
rs752969987 Chr.15: 41504852-41504852 on Build GRCh38
rs371231951 Chr.15: 41504752-41504752 on Build GRCh38
rs755420908 Chr.15: 41504729-41504729 on Build GRCh38
rs370182079 Chr.15: 41504804-41504804 on Build GRCh38

View Full Product Details