Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 73407448-73407721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM168A
Entrez Gene ID
23201
Gene Name
family with sequence similarity 168 member A
Gene Aliases
KIAA0280, TCRP1
UniGene
Hs.475334
Gene Chromosome Location
Chr.11: 73400478-73598189 on Build GRCh38
Transcripts
NM_001286050.1 NM_001286051.1 NM_015159.2 XM_017017415.1 XM_017017416.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6307988 c.680C>A p.A227E Chr.11: 73407559-73407559 on Build GRCh38
7367130 c.655A>G p.T219A Chr.11: 73407584-73407584 on Build GRCh38
320257 c.652C>A p.P218T Chr.11: 73407587-73407587 on Build GRCh38
931816 c.691G>A p.V231M Chr.11: 73407548-73407548 on Build GRCh38
4518480 c.663_664GG>AA p.A222T Chr.11: 73407575-73407576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765935326 Chr.11: 73407694-73407694 on Build GRCh38
rs138819025 Chr.11: 73407695-73407695 on Build GRCh38
rs371613362 Chr.11: 73407571-73407571 on Build GRCh38
rs760764427 Chr.11: 73407482-73407482 on Build GRCh38
rs780140343 Chr.11: 73407577-73407577 on Build GRCh38
rs367570839 Chr.11: 73407613-73407613 on Build GRCh38
rs747946952 Chr.11: 73407602-73407602 on Build GRCh38
rs374821207 Chr.11: 73407584-73407584 on Build GRCh38
rs780968173 Chr.11: 73407565-73407565 on Build GRCh38
rs769799497 Chr.11: 73407616-73407616 on Build GRCh38
rs566820360 Chr.11: 73407605-73407605 on Build GRCh38
rs553310172 Chr.11: 73407590-73407590 on Build GRCh38
rs374037531 Chr.11: 73407549-73407549 on Build GRCh38
rs376655460 Chr.11: 73407659-73407659 on Build GRCh38
rs771195412 Chr.11: 73407610-73407610 on Build GRCh38
rs774625959 Chr.11: 73407661-73407661 on Build GRCh38
rs960626941 Chr.11: 73407499-73407499 on Build GRCh38
rs928240134 Chr.11: 73407498-73407498 on Build GRCh38
rs201384677 Chr.11: 73407558-73407558 on Build GRCh38
rs750008935 Chr.11: 73407570-73407570 on Build GRCh38
rs749118513 Chr.11: 73407582-73407582 on Build GRCh38
rs776734568 Chr.11: 73407501-73407501 on Build GRCh38
rs1010935856 Chr.11: 73407681-73407681 on Build GRCh38
rs567482294 Chr.11: 73407555-73407555 on Build GRCh38
rs765792564 Chr.11: 73407540-73407540 on Build GRCh38
rs78307674 Chr.11: 73407615-73407615 on Build GRCh38
rs776079192 Chr.11: 73407628-73407628 on Build GRCh38
rs755694903 Chr.11: 73407684-73407684 on Build GRCh38
rs756898716 Chr.11: 73407559-73407559 on Build GRCh38
rs371058444 Chr.11: 73407515-73407515 on Build GRCh38
rs773493112 Chr.11: 73407480-73407480 on Build GRCh38
rs559007371 Chr.11: 73407677-73407677 on Build GRCh38
rs762223445 Chr.11: 73407671-73407671 on Build GRCh38
rs767341865 Chr.11: 73407676-73407676 on Build GRCh38
rs752569750 Chr.11: 73407548-73407548 on Build GRCh38
rs981395485 Chr.11: 73407478-73407478 on Build GRCh38
rs780329880 Chr.11: 73407651-73407651 on Build GRCh38
rs759591001 Chr.11: 73407510-73407510 on Build GRCh38
rs771032326 Chr.11: 73407488-73407488 on Build GRCh38

View Full Product Details