Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 869962-870191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHID1
Entrez Gene ID
66005
Gene Name
chitinase domain containing 1
Gene Aliases
GL008, SI-CLP, SICLP
UniGene
Hs.144468
Gene Chromosome Location
Chr.11: 867859-915058 on Build GRCh38
Transcripts
NM_001142674.1 NM_001142675.1 NM_001142676.1 NM_001142677.1 NM_023947.3 XM_005253072.2 XM_005253073.4 XM_006718282.2 XM_011520299.1 XM_011520300.1 XM_017018166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7278238 c.1149C>T p.H383H Chr.11: 870145-870145 on Build GRCh38
5373761 c.1083+1G>A p.? Chr.11: 870120-870120 on Build GRCh38
1492714 c.1055G>C p.R352T Chr.11: 870149-870149 on Build GRCh38
7291877 c.1083+4C>T p.? Chr.11: 870117-870117 on Build GRCh38
5373762 c.1173+1G>A p.? Chr.11: 870120-870120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369487093 Chr.11: 870111-870111 on Build GRCh38
rs751161259 Chr.11: 870116-870116 on Build GRCh38
rs754969137 Chr.11: 869990-869990 on Build GRCh38
rs568144760 Chr.11: 870076-870076 on Build GRCh38
rs748714060 Chr.11: 870144-870144 on Build GRCh38
rs201415316 Chr.11: 870094-870094 on Build GRCh38
rs202154539 Chr.11: 870097-870097 on Build GRCh38
rs957446169 Chr.11: 870027-870027 on Build GRCh38
rs769175892 Chr.11: 870075-870075 on Build GRCh38
rs752274383 Chr.11: 870000-870000 on Build GRCh38
rs138166915 Chr.11: 870141-870141 on Build GRCh38
rs370593417 Chr.11: 870156-870156 on Build GRCh38
rs905764501 Chr.11: 870142-870142 on Build GRCh38
rs200188854 Chr.11: 870001-870001 on Build GRCh38
rs113580509 Chr.11: 870095-870095 on Build GRCh38
rs773470775 Chr.11: 870088-870088 on Build GRCh38
rs141753287 Chr.11: 870095-870095 on Build GRCh38
rs1044084682 Chr.11: 870084-870084 on Build GRCh38
rs770175300 Chr.11: 870041-870041 on Build GRCh38
rs754840899 Chr.11: 870117-870117 on Build GRCh38
rs115994760 Chr.11: 870136-870136 on Build GRCh38
rs562937742 Chr.11: 870031-870031 on Build GRCh38
rs746685245 Chr.11: 870004-870004 on Build GRCh38
rs980054010 Chr.11: 870013-870013 on Build GRCh38
rs140476082 Chr.11: 870158-870158 on Build GRCh38
rs1032977173 Chr.11: 870148-870148 on Build GRCh38
rs770193297 Chr.11: 870006-870006 on Build GRCh38
rs948349166 Chr.11: 870081-870081 on Build GRCh38
rs201931075 Chr.11: 870074-870074 on Build GRCh38
rs886359843 Chr.11: 869989-869989 on Build GRCh38
rs1047934105 Chr.11: 870115-870115 on Build GRCh38
rs560853222 Chr.11: 870011-870011 on Build GRCh38
rs776443991 Chr.11: 870166-870166 on Build GRCh38
rs144295528 Chr.11: 870154-870154 on Build GRCh38
rs759497130 Chr.11: 870102-870102 on Build GRCh38
rs780419403 Chr.11: 870120-870120 on Build GRCh38
rs769049099 Chr.11: 870137-870137 on Build GRCh38
rs376509515 Chr.11: 870087-870087 on Build GRCh38
rs150184057 Chr.11: 870028-870028 on Build GRCh38
rs780290124 Chr.11: 870073-870073 on Build GRCh38
rs115326504 Chr.11: 870040-870040 on Build GRCh38
rs987080839 Chr.11: 870072-870072 on Build GRCh38
rs748378823 Chr.11: 870090-870090 on Build GRCh38
rs532653739 Chr.11: 870015-870015 on Build GRCh38
rs570735805 Chr.11: 870103-870103 on Build GRCh38
rs573982321 Chr.11: 869998-869998 on Build GRCh38
rs372856810 Chr.11: 869988-869988 on Build GRCh38
rs373291423 Chr.11: 870077-870077 on Build GRCh38
rs149585062 Chr.11: 870145-870145 on Build GRCh38

View Full Product Details