Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154571941-154572166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNB2
Entrez Gene ID
1141
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Gene Aliases
EFNL3, nAChRB2
UniGene
Hs.2306
Gene Chromosome Location
Chr.1: 154567781-154580026 on Build GRCh38
Transcripts
NM_000748.2 XM_017000180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4022831 c.1137C>A p.R379R Chr.1: 154571960-154571960 on Build GRCh38
6575876 c.1252G>A p.E418K Chr.1: 154572075-154572075 on Build GRCh38
4140850 c.1151C>T p.A384V Chr.1: 154571974-154571974 on Build GRCh38
3771464 c.1190A>G p.Q397R Chr.1: 154572013-154572013 on Build GRCh38
3771463 c.1137C>T p.R379R Chr.1: 154571960-154571960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767533378 Chr.1: 154572059-154572059 on Build GRCh38
rs770055798 Chr.1: 154571994-154571994 on Build GRCh38
rs35834206 Chr.1: 154572013-154572014 on Build GRCh38
rs372757638 Chr.1: 154572062-154572062 on Build GRCh38
rs746495548 Chr.1: 154571975-154571975 on Build GRCh38
rs796052331 Chr.1: 154571994-154572003 on Build GRCh38
rs748619028 Chr.1: 154571983-154571983 on Build GRCh38
rs758068217 Chr.1: 154571965-154571965 on Build GRCh38
rs560427247 Chr.1: 154571999-154571999 on Build GRCh38
rs77710036 Chr.1: 154572096-154572096 on Build GRCh38
rs763142542 Chr.1: 154572055-154572055 on Build GRCh38
rs902984484 Chr.1: 154572024-154572024 on Build GRCh38
rs774449832 Chr.1: 154572057-154572057 on Build GRCh38
rs771273497 Chr.1: 154572139-154572139 on Build GRCh38
rs971173003 Chr.1: 154572067-154572067 on Build GRCh38
rs1038602573 Chr.1: 154571960-154571960 on Build GRCh38
rs1024048381 Chr.1: 154572063-154572063 on Build GRCh38
rs974346112 Chr.1: 154572081-154572081 on Build GRCh38
rs996179072 Chr.1: 154571995-154571995 on Build GRCh38
rs990542717 Chr.1: 154572133-154572133 on Build GRCh38
rs200910048 Chr.1: 154572137-154572137 on Build GRCh38
rs199658508 Chr.1: 154572027-154572027 on Build GRCh38
rs200441832 Chr.1: 154572013-154572013 on Build GRCh38
rs1020953003 Chr.1: 154572022-154572022 on Build GRCh38
rs1020120934 Chr.1: 154572023-154572023 on Build GRCh38
rs55857552 Chr.1: 154572056-154572056 on Build GRCh38
rs747248135 Chr.1: 154572118-154572118 on Build GRCh38
rs901932970 Chr.1: 154571967-154571967 on Build GRCh38
rs942920445 Chr.1: 154572037-154572048 on Build GRCh38
rs76126473 Chr.1: 154572089-154572089 on Build GRCh38
rs55685423 Chr.1: 154572014-154572014 on Build GRCh38
rs542041934 Chr.1: 154571980-154571980 on Build GRCh38
rs112585933 Chr.1: 154572058-154572058 on Build GRCh38
rs908723251 Chr.1: 154572079-154572079 on Build GRCh38
rs1049048240 Chr.1: 154572143-154572143 on Build GRCh38
rs932622161 Chr.1: 154571976-154571976 on Build GRCh38
rs527768017 Chr.1: 154572001-154572001 on Build GRCh38
rs201986342 Chr.1: 154571969-154571969 on Build GRCh38
rs753784853 Chr.1: 154572122-154572122 on Build GRCh38
rs779771980 Chr.1: 154571972-154571972 on Build GRCh38
rs199743038 Chr.1: 154572080-154572080 on Build GRCh38
rs945492622 Chr.1: 154572138-154572138 on Build GRCh38
rs982906227 Chr.1: 154572077-154572077 on Build GRCh38
rs916448473 Chr.1: 154572135-154572135 on Build GRCh38

View Full Product Details