Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31539080-31539343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1G2-DDX39B
Entrez Gene ID
100532737
Gene Name
ATP6V1G2-DDX39B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.254042
Gene Chromosome Location
Chr.6: 31530219-31546848 on Build GRCh38
Transcripts
NR_037853.1 NM_004640.6 NM_080598.5 NR_037852.1

Gene Symbol
DDX39B
Entrez Gene ID
7919
Gene Name
DEAD-box helicase 39B
Gene Aliases
BAT1, D6S81E, UAP56
UniGene
Hs.254042
Gene Chromosome Location
Chr.6: 31530219-31542475 on Build GRCh38
Transcripts
NR_037853.1 NM_004640.6 NM_080598.5 NR_037852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4676819 c.257T>C p.L86P Chr.6: 31539229-31539229 on Build GRCh38
7196027 c.349_350insG p.T119fs*39 Chr.6: 31539205-31539206 on Build GRCh38
4857250 c.239T>C p.I80T Chr.6: 31539247-31539247 on Build GRCh38
4676821 c.257T>C p.L86P Chr.6: 31539229-31539229 on Build GRCh38
7196024 c.280_281insG p.T96fs*39 Chr.6: 31539205-31539206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768125874 Chr.6: 31539284-31539284 on Build GRCh38
rs879229025 Chr.6: 31539119-31539119 on Build GRCh38
rs756521014 Chr.6: 31539264-31539264 on Build GRCh38
rs775195232 Chr.6: 31539108-31539108 on Build GRCh38
rs758717148 Chr.6: 31539286-31539286 on Build GRCh38
rs371761523 Chr.6: 31539287-31539287 on Build GRCh38
rs1032432098 Chr.6: 31539219-31539219 on Build GRCh38
rs2071595 Chr.6: 31539285-31539285 on Build GRCh38
rs751909909 Chr.6: 31539297-31539297 on Build GRCh38
rs772529969 Chr.6: 31539301-31539301 on Build GRCh38
rs762764612 Chr.6: 31539112-31539112 on Build GRCh38
rs766929998 Chr.6: 31539158-31539158 on Build GRCh38
rs148931659 Chr.6: 31539278-31539278 on Build GRCh38
rs746273190 Chr.6: 31539283-31539284 on Build GRCh38
rs920470241 Chr.6: 31539213-31539213 on Build GRCh38
rs754216255 Chr.6: 31539283-31539283 on Build GRCh38
rs773967863 Chr.6: 31539142-31539142 on Build GRCh38
rs62395764 Chr.6: 31539280-31539280 on Build GRCh38
rs767211584 Chr.6: 31539298-31539298 on Build GRCh38
rs768406725 Chr.6: 31539292-31539292 on Build GRCh38
rs781487959 Chr.6: 31539282-31539283 on Build GRCh38
rs774041679 Chr.6: 31539292-31539292 on Build GRCh38
rs183581681 Chr.6: 31539290-31539290 on Build GRCh38
rs187497360 Chr.6: 31539294-31539294 on Build GRCh38
rs763684827 Chr.6: 31539140-31539140 on Build GRCh38
rs150991114 Chr.6: 31539153-31539153 on Build GRCh38
rs775426504 Chr.6: 31539288-31539288 on Build GRCh38
rs760264014 Chr.6: 31539309-31539309 on Build GRCh38
rs750007290 Chr.6: 31539167-31539167 on Build GRCh38

View Full Product Details