Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49793212-49793439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDHR4
Entrez Gene ID
389118
Gene Name
cadherin related family member 4
Gene Aliases
CDH29, PRO34300
UniGene
Hs.363312
Gene Chromosome Location
Chr.3: 49790732-49802741 on Build GRCh38
Transcripts
NM_001007540.3 XM_011533700.2 XM_011533701.2 XM_017006366.1 XM_017006367.1 XM_017006368.1 XM_017006369.1 XM_017006370.1 XM_017006371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5185011 c.1694G>A p.R565H Chr.3: 49793241-49793241 on Build GRCh38
5984774 c.1707G>A p.V569V Chr.3: 49793228-49793228 on Build GRCh38
4471275 c.1709C>T p.T570I Chr.3: 49793226-49793226 on Build GRCh38
5599277 c.1677C>G p.I559M Chr.3: 49793258-49793258 on Build GRCh38
6584352 c.1663C>G p.Q555E Chr.3: 49793272-49793272 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs912644931 Chr.3: 49793233-49793233 on Build GRCh38
rs1029792092 Chr.3: 49793241-49793241 on Build GRCh38
rs775360966 Chr.3: 49793417-49793417 on Build GRCh38
rs777125796 Chr.3: 49793287-49793287 on Build GRCh38
rs1037122507 Chr.3: 49793256-49793256 on Build GRCh38
rs753507796 Chr.3: 49793359-49793359 on Build GRCh38
rs902523266 Chr.3: 49793235-49793235 on Build GRCh38
rs755832689 Chr.3: 49793276-49793276 on Build GRCh38
rs954684013 Chr.3: 49793272-49793272 on Build GRCh38
rs141327397 Chr.3: 49793362-49793362 on Build GRCh38
rs973732284 Chr.3: 49793295-49793295 on Build GRCh38
rs559952714 Chr.3: 49793290-49793290 on Build GRCh38
rs148137199 Chr.3: 49793236-49793236 on Build GRCh38
rs983143393 Chr.3: 49793397-49793397 on Build GRCh38
rs950423687 Chr.3: 49793378-49793378 on Build GRCh38
rs964637519 Chr.3: 49793293-49793293 on Build GRCh38
rs768700230 Chr.3: 49793357-49793357 on Build GRCh38
rs999603184 Chr.3: 49793237-49793237 on Build GRCh38
rs541668125 Chr.3: 49793262-49793262 on Build GRCh38
rs113429839 Chr.3: 49793396-49793397 on Build GRCh38
rs1055432158 Chr.3: 49793338-49793338 on Build GRCh38
rs868117646 Chr.3: 49793274-49793274 on Build GRCh38
rs368005921 Chr.3: 49793291-49793291 on Build GRCh38

View Full Product Details