Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 54811213-54811442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGED2
Entrez Gene ID
10916
Gene Name
MAGE family member D2
Gene Aliases
11B6, BARTS5, BCG-1, BCG1, HCA10, MAGE-D2
UniGene
Hs.522665
Gene Chromosome Location
Chr.X: 54807599-54816015 on Build GRCh38
Transcripts
NM_014599.5 NM_177433.2 NM_201222.2

SNP Detail

SNP ID Chromosome Location
rs755292405 Chr.X: 54811295-54811295 on Build GRCh38
rs759292577 Chr.X: 54811359-54811359 on Build GRCh38
rs138036392 Chr.X: 54811290-54811290 on Build GRCh38
rs781562704 Chr.X: 54811296-54811296 on Build GRCh38
rs774247830 Chr.X: 54811358-54811358 on Build GRCh38
rs749078330 Chr.X: 54811300-54811300 on Build GRCh38
rs900145962 Chr.X: 54811377-54811377 on Build GRCh38
rs138103264 Chr.X: 54811331-54811331 on Build GRCh38
rs1040044230 Chr.X: 54811373-54811373 on Build GRCh38
rs764365185 Chr.X: 54811370-54811370 on Build GRCh38
rs771039734 Chr.X: 54811312-54811312 on Build GRCh38
rs770721700 Chr.X: 54811333-54811333 on Build GRCh38
rs372339836 Chr.X: 54811287-54811287 on Build GRCh38
rs770006492 Chr.X: 54811318-54811318 on Build GRCh38
rs758795312 Chr.X: 54811247-54811247 on Build GRCh38
rs920335549 Chr.X: 54811372-54811372 on Build GRCh38
rs750948304 Chr.X: 54811245-54811245 on Build GRCh38
rs375727148 Chr.X: 54811328-54811328 on Build GRCh38
rs754040319 Chr.X: 54811414-54811414 on Build GRCh38
rs767222403 Chr.X: 54811362-54811362 on Build GRCh38
rs372424163 Chr.X: 54811334-54811334 on Build GRCh38

View Full Product Details