Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 22388147-22388390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRA3
Entrez Gene ID
166647
Gene Name
adhesion G protein-coupled receptor A3
Gene Aliases
GPR125, PGR21, TEM5L
UniGene
Hs.684429
Gene Chromosome Location
Chr.4: 22387374-22516058 on Build GRCh38
Transcripts
NM_145290.3 XM_011513811.2 XM_017007834.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4687956 c.3498G>A p.V1166V Chr.4: 22388173-22388173 on Build GRCh38
399140 c.3310T>A p.S1104T Chr.4: 22388361-22388361 on Build GRCh38
1054534 c.3282G>A p.A1094A Chr.4: 22388389-22388389 on Build GRCh38
5569098 c.3428C>G p.T1143R Chr.4: 22388243-22388243 on Build GRCh38
733705 c.3513C>T p.H1171H Chr.4: 22388158-22388158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972347319 Chr.4: 22388267-22388267 on Build GRCh38
rs769898786 Chr.4: 22388285-22388285 on Build GRCh38
rs947300211 Chr.4: 22388328-22388328 on Build GRCh38
rs769226062 Chr.4: 22388293-22388293 on Build GRCh38
rs781447096 Chr.4: 22388344-22388344 on Build GRCh38
rs750684538 Chr.4: 22388349-22388349 on Build GRCh38
rs199616944 Chr.4: 22388193-22388193 on Build GRCh38
rs756220717 Chr.4: 22388360-22388360 on Build GRCh38
rs777549706 Chr.4: 22388266-22388266 on Build GRCh38
rs922253997 Chr.4: 22388250-22388250 on Build GRCh38
rs763279377 Chr.4: 22388208-22388208 on Build GRCh38
rs371121360 Chr.4: 22388303-22388303 on Build GRCh38
rs879876606 Chr.4: 22388280-22388280 on Build GRCh38
rs764007687 Chr.4: 22388337-22388337 on Build GRCh38
rs143592195 Chr.4: 22388244-22388244 on Build GRCh38
rs148226840 Chr.4: 22388203-22388203 on Build GRCh38
rs776038380 Chr.4: 22388204-22388204 on Build GRCh38
rs774785929 Chr.4: 22388212-22388212 on Build GRCh38
rs771076973 Chr.4: 22388183-22388183 on Build GRCh38
rs753463580 Chr.4: 22388222-22388222 on Build GRCh38
rs757536970 Chr.4: 22388271-22388271 on Build GRCh38
rs189292382 Chr.4: 22388191-22388191 on Build GRCh38
rs765136152 Chr.4: 22388235-22388235 on Build GRCh38
rs545763677 Chr.4: 22388288-22388288 on Build GRCh38
rs751299637 Chr.4: 22388270-22388270 on Build GRCh38
rs752733441 Chr.4: 22388177-22388177 on Build GRCh38
rs757017927 Chr.4: 22388342-22388342 on Build GRCh38
rs762262067 Chr.4: 22388301-22388301 on Build GRCh38
rs766112442 Chr.4: 22388217-22388217 on Build GRCh38
rs201521604 Chr.4: 22388213-22388213 on Build GRCh38
rs751426456 Chr.4: 22388341-22388341 on Build GRCh38
rs764835658 Chr.4: 22388312-22388312 on Build GRCh38
rs909429953 Chr.4: 22388231-22388231 on Build GRCh38
rs111298499 Chr.4: 22388365-22388365 on Build GRCh38
rs755572190 Chr.4: 22388286-22388286 on Build GRCh38
rs138052583 Chr.4: 22388223-22388223 on Build GRCh38
rs3814416 Chr.4: 22388174-22388174 on Build GRCh38
rs749313362 Chr.4: 22388367-22388367 on Build GRCh38
rs774910758 Chr.4: 22388294-22388294 on Build GRCh38
rs201813100 Chr.4: 22388168-22388168 on Build GRCh38
rs564337647 Chr.4: 22388311-22388311 on Build GRCh38
rs574152051 Chr.4: 22388209-22388209 on Build GRCh38
rs1035873766 Chr.4: 22388202-22388202 on Build GRCh38
rs557122010 Chr.4: 22388358-22388358 on Build GRCh38
rs200450492 Chr.4: 22388243-22388243 on Build GRCh38
rs776705989 Chr.4: 22388188-22388188 on Build GRCh38
rs755147070 Chr.4: 22388282-22388282 on Build GRCh38

View Full Product Details