Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 61766328-61766559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PATJ
Entrez Gene ID
10207
Gene Name
PATJ, crumbs cell polarity complex component
Gene Aliases
Cipp, INADL, InaD-like, hINADL
UniGene
Hs.478125
Gene Chromosome Location
Chr.1: 61742477-62163920 on Build GRCh38
Transcripts
NM_176877.2 XM_005270341.3 XM_005270347.1 XM_006710278.3 XM_011540462.2 XM_011540463.2 XM_011540464.2 XM_011540465.2 XM_011540466.2 XM_011540467.2 XM_011540468.2 XM_011540469.2 XM_011540470.1 XM_016999998.1 XM_016999999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6685672 c.336G>A p.P112P Chr.1: 61766425-61766425 on Build GRCh38
1343747 c.311T>C p.V104A Chr.1: 61766400-61766400 on Build GRCh38
4825126 c.352G>A p.D118N Chr.1: 61766441-61766441 on Build GRCh38
5425301 c.369T>C p.I123I Chr.1: 61766458-61766458 on Build GRCh38
281926 c.306G>A p.S102S Chr.1: 61766395-61766395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779407233 Chr.1: 61766424-61766424 on Build GRCh38
rs372320578 Chr.1: 61766466-61766466 on Build GRCh38
rs774360860 Chr.1: 61766379-61766379 on Build GRCh38
rs760918830 Chr.1: 61766395-61766395 on Build GRCh38
rs151215734 Chr.1: 61766415-61766415 on Build GRCh38
rs779773939 Chr.1: 61766484-61766484 on Build GRCh38
rs753912855 Chr.1: 61766416-61766416 on Build GRCh38
rs761709861 Chr.1: 61766383-61766383 on Build GRCh38
rs868427975 Chr.1: 61766460-61766460 on Build GRCh38
rs753154796 Chr.1: 61766428-61766428 on Build GRCh38
rs954990209 Chr.1: 61766425-61766425 on Build GRCh38
rs11207827 Chr.1: 61766458-61766458 on Build GRCh38
rs557978080 Chr.1: 61766514-61766514 on Build GRCh38
rs987570635 Chr.1: 61766446-61766446 on Build GRCh38
rs774448740 Chr.1: 61766524-61766524 on Build GRCh38
rs754986630 Chr.1: 61766422-61766422 on Build GRCh38
rs190078763 Chr.1: 61766394-61766394 on Build GRCh38
rs1290444 Chr.1: 61766501-61766501 on Build GRCh38
rs767290853 Chr.1: 61766388-61766388 on Build GRCh38
rs902912635 Chr.1: 61766472-61766472 on Build GRCh38
rs1286835 Chr.1: 61766359-61766359 on Build GRCh38
rs758725472 Chr.1: 61766449-61766449 on Build GRCh38
rs183271737 Chr.1: 61766482-61766482 on Build GRCh38

View Full Product Details