Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84563256-84563481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTBS
Entrez Gene ID
1486
Gene Name
chitobiase
Gene Aliases
CTB
UniGene
Hs.513557
Gene Chromosome Location
Chr.1: 84553121-84574480 on Build GRCh38
Transcripts
NM_004388.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4510001 c.823C>T p.P275S Chr.1: 84563391-84563391 on Build GRCh38
6230320 c.878C>T p.T293M Chr.1: 84563336-84563336 on Build GRCh38
912585 c.879G>A p.T293T Chr.1: 84563335-84563335 on Build GRCh38
912586 c.860G>A p.R287H Chr.1: 84563354-84563354 on Build GRCh38
681915 c.830G>T p.R277L Chr.1: 84563384-84563384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541565613 Chr.1: 84563366-84563366 on Build GRCh38
rs369074009 Chr.1: 84563403-84563403 on Build GRCh38
rs955542700 Chr.1: 84563369-84563369 on Build GRCh38
rs770404895 Chr.1: 84563395-84563395 on Build GRCh38
rs758636345 Chr.1: 84563378-84563379 on Build GRCh38
rs145768666 Chr.1: 84563376-84563376 on Build GRCh38
rs555313581 Chr.1: 84563437-84563439 on Build GRCh38
rs938480921 Chr.1: 84563326-84563326 on Build GRCh38
rs764489979 Chr.1: 84563421-84563421 on Build GRCh38
rs77922958 Chr.1: 84563355-84563355 on Build GRCh38
rs759361175 Chr.1: 84563360-84563360 on Build GRCh38
rs776020910 Chr.1: 84563416-84563416 on Build GRCh38
rs375604956 Chr.1: 84563336-84563336 on Build GRCh38
rs781330409 Chr.1: 84563379-84563379 on Build GRCh38
rs765879098 Chr.1: 84563347-84563347 on Build GRCh38
rs779907493 Chr.1: 84563304-84563304 on Build GRCh38
rs780187443 Chr.1: 84563429-84563429 on Build GRCh38
rs1039771293 Chr.1: 84563288-84563288 on Build GRCh38
rs372939098 Chr.1: 84563344-84563344 on Build GRCh38
rs764296657 Chr.1: 84563407-84563407 on Build GRCh38
rs148118320 Chr.1: 84563384-84563384 on Build GRCh38
rs367692581 Chr.1: 84563365-84563365 on Build GRCh38
rs138752249 Chr.1: 84563385-84563385 on Build GRCh38
rs376746415 Chr.1: 84563315-84563315 on Build GRCh38
rs373912484 Chr.1: 84563350-84563350 on Build GRCh38
rs751797912 Chr.1: 84563428-84563428 on Build GRCh38
rs879025129 Chr.1: 84563383-84563383 on Build GRCh38
rs772777295 Chr.1: 84563392-84563392 on Build GRCh38
rs761472620 Chr.1: 84563324-84563324 on Build GRCh38
rs749193629 Chr.1: 84563306-84563306 on Build GRCh38
rs35270754 Chr.1: 84563398-84563398 on Build GRCh38
rs754461413 Chr.1: 84563349-84563349 on Build GRCh38
rs1055940606 Chr.1: 84563335-84563335 on Build GRCh38
rs747885910 Chr.1: 84563386-84563386 on Build GRCh38
rs757491234 Chr.1: 84563433-84563433 on Build GRCh38
rs771875024 Chr.1: 84563390-84563390 on Build GRCh38
rs772796713 Chr.1: 84563331-84563331 on Build GRCh38
rs201997781 Chr.1: 84563327-84563327 on Build GRCh38
rs374103195 Chr.1: 84563364-84563364 on Build GRCh38
rs755345734 Chr.1: 84563452-84563453 on Build GRCh38
rs143993403 Chr.1: 84563402-84563402 on Build GRCh38
rs928283673 Chr.1: 84563318-84563318 on Build GRCh38
rs15911 Chr.1: 84563394-84563394 on Build GRCh38
rs774660684 Chr.1: 84563310-84563310 on Build GRCh38
rs201232040 Chr.1: 84563417-84563417 on Build GRCh38
rs3768249 Chr.1: 84563286-84563286 on Build GRCh38
rs375887489 Chr.1: 84563354-84563354 on Build GRCh38
rs987027323 Chr.1: 84563415-84563415 on Build GRCh38

View Full Product Details