Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20806847-20807106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC81691
Entrez Gene ID
81691
Gene Name
exonuclease NEF-sp
Gene Aliases
-
UniGene
Hs.177926
Gene Chromosome Location
Chr.16: 20806429-20849668 on Build GRCh38
Transcripts
NM_001144924.1 NM_001199053.1 NM_030941.2 XM_005255604.1 XM_011545963.1 XM_011545964.1 XM_011545965.1 XM_011545966.1 XM_017023739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5363318 c.50A>G p.E17G Chr.16: 20807003-20807003 on Build GRCh38
4882162 c.130G>A p.E44K Chr.16: 20807083-20807083 on Build GRCh38
5983297 c.21G>A p.G7G Chr.16: 20806974-20806974 on Build GRCh38
5552141 c.110A>G p.D37G Chr.16: 20807063-20807063 on Build GRCh38
968166 c.134C>A p.A45D Chr.16: 20807087-20807087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953138823 Chr.16: 20806923-20806923 on Build GRCh38
rs745451778 Chr.16: 20807068-20807068 on Build GRCh38
rs756589714 Chr.16: 20807036-20807036 on Build GRCh38
rs746692862 Chr.16: 20807084-20807084 on Build GRCh38
rs755146835 Chr.16: 20806995-20806995 on Build GRCh38
rs1030925928 Chr.16: 20806901-20806901 on Build GRCh38
rs941096826 Chr.16: 20806896-20806896 on Build GRCh38
rs999224251 Chr.16: 20806898-20806898 on Build GRCh38
rs963077036 Chr.16: 20806980-20806980 on Build GRCh38
rs1017679216 Chr.16: 20807021-20807021 on Build GRCh38
rs61737146 Chr.16: 20807067-20807067 on Build GRCh38
rs964813181 Chr.16: 20807023-20807023 on Build GRCh38
rs775174603 Chr.16: 20807080-20807080 on Build GRCh38
rs781619658 Chr.16: 20807020-20807020 on Build GRCh38
rs190537811 Chr.16: 20806897-20806897 on Build GRCh38
rs985893583 Chr.16: 20806926-20806926 on Build GRCh38
rs751660395 Chr.16: 20806943-20806943 on Build GRCh38
rs1009746897 Chr.16: 20806949-20806949 on Build GRCh38
rs141233630 Chr.16: 20807046-20807046 on Build GRCh38
rs766311218 Chr.16: 20806924-20806924 on Build GRCh38
rs892360572 Chr.16: 20806910-20806910 on Build GRCh38
rs903827165 Chr.16: 20807000-20807000 on Build GRCh38
rs372123593 Chr.16: 20806919-20806919 on Build GRCh38
rs753087699 Chr.16: 20807035-20807035 on Build GRCh38
rs370157102 Chr.16: 20806913-20806913 on Build GRCh38
rs938211294 Chr.16: 20806905-20806905 on Build GRCh38
rs771611246 Chr.16: 20807073-20807073 on Build GRCh38
rs908176883 Chr.16: 20806892-20806892 on Build GRCh38
rs1016381414 Chr.16: 20806961-20806961 on Build GRCh38
rs200945676 Chr.16: 20806954-20806954 on Build GRCh38
rs374764285 Chr.16: 20807052-20807052 on Build GRCh38
rs200638604 Chr.16: 20807086-20807086 on Build GRCh38
rs904937595 Chr.16: 20806899-20806899 on Build GRCh38
rs1042378360 Chr.16: 20806987-20806987 on Build GRCh38
rs201747673 Chr.16: 20807058-20807058 on Build GRCh38

View Full Product Details