Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 53823052-53823297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF2B
Entrez Gene ID
84643
Gene Name
kinesin family member 2B
Gene Aliases
-
UniGene
Hs.226805
Gene Chromosome Location
Chr.17: 53822878-53825212 on Build GRCh38
Transcripts
NM_032559.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6199507 c.44C>T p.P15L Chr.17: 53823077-53823077 on Build GRCh38
6556493 c.90C>T p.I30I Chr.17: 53823123-53823123 on Build GRCh38
6147218 c.45C>A p.P15P Chr.17: 53823078-53823078 on Build GRCh38
3519656 c.183G>A p.V61V Chr.17: 53823216-53823216 on Build GRCh38
6147216 c.204C>A p.G68G Chr.17: 53823237-53823237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760154206 Chr.17: 53823238-53823240 on Build GRCh38
rs375544135 Chr.17: 53823116-53823116 on Build GRCh38
rs777744072 Chr.17: 53823102-53823102 on Build GRCh38
rs566108739 Chr.17: 53823074-53823074 on Build GRCh38
rs375840279 Chr.17: 53823195-53823195 on Build GRCh38
rs754482684 Chr.17: 53823140-53823140 on Build GRCh38
rs773748663 Chr.17: 53823236-53823236 on Build GRCh38
rs554238256 Chr.17: 53823266-53823266 on Build GRCh38
rs756888791 Chr.17: 53823269-53823269 on Build GRCh38
rs527417913 Chr.17: 53823075-53823075 on Build GRCh38
rs778866434 Chr.17: 53823174-53823174 on Build GRCh38
rs762229162 Chr.17: 53823229-53823229 on Build GRCh38
rs969936039 Chr.17: 53823119-53823119 on Build GRCh38
rs777158812 Chr.17: 53823208-53823208 on Build GRCh38
rs999738557 Chr.17: 53823263-53823263 on Build GRCh38
rs7216557 Chr.17: 53823072-53823072 on Build GRCh38
rs771625502 Chr.17: 53823076-53823076 on Build GRCh38
rs114564922 Chr.17: 53823132-53823132 on Build GRCh38
rs772212177 Chr.17: 53823125-53823125 on Build GRCh38
rs754609985 Chr.17: 53823073-53823073 on Build GRCh38
rs767122162 Chr.17: 53823151-53823151 on Build GRCh38
rs757160514 Chr.17: 53823173-53823173 on Build GRCh38
rs745907431 Chr.17: 53823107-53823107 on Build GRCh38
rs770787002 Chr.17: 53823103-53823103 on Build GRCh38
rs764552763 Chr.17: 53823128-53823128 on Build GRCh38
rs763682994 Chr.17: 53823273-53823273 on Build GRCh38
rs761182386 Chr.17: 53823127-53823127 on Build GRCh38
rs768946124 Chr.17: 53823204-53823204 on Build GRCh38
rs944869719 Chr.17: 53823118-53823118 on Build GRCh38
rs767047890 Chr.17: 53823260-53823260 on Build GRCh38
rs941205329 Chr.17: 53823145-53823145 on Build GRCh38
rs201197805 Chr.17: 53823207-53823207 on Build GRCh38
rs889468547 Chr.17: 53823210-53823210 on Build GRCh38
rs775054535 Chr.17: 53823118-53823128 on Build GRCh38
rs749020826 Chr.17: 53823169-53823169 on Build GRCh38
rs755879983 Chr.17: 53823078-53823078 on Build GRCh38
rs369661767 Chr.17: 53823135-53823135 on Build GRCh38
rs551845820 Chr.17: 53823077-53823077 on Build GRCh38
rs369270831 Chr.17: 53823233-53823233 on Build GRCh38
rs747241718 Chr.17: 53823198-53823198 on Build GRCh38
rs971647972 Chr.17: 53823228-53823229 on Build GRCh38
rs775762338 Chr.17: 53823126-53823126 on Build GRCh38
rs752134865 Chr.17: 53823262-53823262 on Build GRCh38
rs777548098 Chr.17: 53823165-53823165 on Build GRCh38
rs1040618464 Chr.17: 53823120-53823120 on Build GRCh38
rs115513389 Chr.17: 53823277-53823277 on Build GRCh38
rs759023753 Chr.17: 53823251-53823251 on Build GRCh38
rs138727670 Chr.17: 53823139-53823139 on Build GRCh38
rs201736778 Chr.17: 53823152-53823152 on Build GRCh38
rs373169435 Chr.17: 53823175-53823175 on Build GRCh38
rs755969751 Chr.17: 53823163-53823163 on Build GRCh38

View Full Product Details