Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45629479-45629752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPBP1L1
Entrez Gene ID
60313
Gene Name
GC-rich promoter binding protein 1 like 1
Gene Aliases
SP192
UniGene
Hs.238432
Gene Chromosome Location
Chr.1: 45627304-45686691 on Build GRCh38
Transcripts
NM_021639.4 XM_017002019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4402095 c.1214A>G p.E405G Chr.1: 45629634-45629634 on Build GRCh38
1343074 c.1213G>A p.E405K Chr.1: 45629635-45629635 on Build GRCh38
7122003 c.1248A>G p.K416K Chr.1: 45629600-45629600 on Build GRCh38
7369828 c.1249G>C p.E417Q Chr.1: 45629599-45629599 on Build GRCh38
4008534 c.1199A>G p.Y400C Chr.1: 45629649-45629649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141695515 Chr.1: 45629610-45629610 on Build GRCh38
rs369188643 Chr.1: 45629691-45629691 on Build GRCh38
rs756914574 Chr.1: 45629631-45629631 on Build GRCh38
rs569434594 Chr.1: 45629687-45629687 on Build GRCh38
rs762034392 Chr.1: 45629573-45629573 on Build GRCh38
rs748882235 Chr.1: 45629626-45629626 on Build GRCh38
rs367608337 Chr.1: 45629616-45629616 on Build GRCh38
rs970810801 Chr.1: 45629536-45629536 on Build GRCh38
rs757814279 Chr.1: 45629730-45629730 on Build GRCh38
rs748584802 Chr.1: 45629678-45629678 on Build GRCh38
rs753622626 Chr.1: 45629699-45629699 on Build GRCh38
rs774441567 Chr.1: 45629685-45629685 on Build GRCh38
rs745373733 Chr.1: 45629727-45629728 on Build GRCh38
rs773538182 Chr.1: 45629683-45629683 on Build GRCh38
rs372107730 Chr.1: 45629625-45629625 on Build GRCh38
rs770047911 Chr.1: 45629681-45629681 on Build GRCh38
rs138133331 Chr.1: 45629593-45629593 on Build GRCh38
rs745331921 Chr.1: 45629660-45629660 on Build GRCh38
rs752590277 Chr.1: 45629620-45629620 on Build GRCh38
rs756704814 Chr.1: 45629538-45629539 on Build GRCh38
rs769309860 Chr.1: 45629665-45629665 on Build GRCh38
rs865930391 Chr.1: 45629609-45629609 on Build GRCh38
rs756260793 Chr.1: 45629713-45629713 on Build GRCh38
rs1025491992 Chr.1: 45629510-45629510 on Build GRCh38
rs763172485 Chr.1: 45629684-45629684 on Build GRCh38
rs148045088 Chr.1: 45629622-45629622 on Build GRCh38
rs376335212 Chr.1: 45629709-45629709 on Build GRCh38
rs778572363 Chr.1: 45629653-45629653 on Build GRCh38
rs374946861 Chr.1: 45629601-45629601 on Build GRCh38
rs763085830 Chr.1: 45629590-45629590 on Build GRCh38
rs747533755 Chr.1: 45629553-45629553 on Build GRCh38
rs376923638 Chr.1: 45629688-45629688 on Build GRCh38
rs1043864345 Chr.1: 45629522-45629522 on Build GRCh38
rs1135812 Chr.1: 45629600-45629600 on Build GRCh38
rs748699935 Chr.1: 45629537-45629538 on Build GRCh38
rs778551561 Chr.1: 45629556-45629558 on Build GRCh38
rs76932789 Chr.1: 45629633-45629633 on Build GRCh38
rs555860388 Chr.1: 45629540-45629540 on Build GRCh38
rs777272948 Chr.1: 45629537-45629537 on Build GRCh38
rs767534259 Chr.1: 45629689-45629689 on Build GRCh38
rs536043469 Chr.1: 45629731-45629731 on Build GRCh38
rs765543554 Chr.1: 45629577-45629577 on Build GRCh38
rs776980680 Chr.1: 45629673-45629673 on Build GRCh38
rs867881022 Chr.1: 45629646-45629646 on Build GRCh38
rs766430945 Chr.1: 45629594-45629594 on Build GRCh38
rs769220097 Chr.1: 45629563-45629563 on Build GRCh38
rs778660259 Chr.1: 45629712-45629712 on Build GRCh38
rs773273845 Chr.1: 45629589-45629589 on Build GRCh38
rs776199346 Chr.1: 45629548-45629548 on Build GRCh38
rs374260191 Chr.1: 45629695-45629695 on Build GRCh38
rs770563518 Chr.1: 45629547-45629547 on Build GRCh38
rs1012422749 Chr.1: 45629602-45629602 on Build GRCh38
rs757002474 Chr.1: 45629702-45629702 on Build GRCh38
rs369910140 Chr.1: 45629725-45629725 on Build GRCh38
rs777085908 Chr.1: 45629566-45629566 on Build GRCh38
rs199675776 Chr.1: 45629694-45629694 on Build GRCh38
rs755818430 Chr.1: 45629535-45629535 on Build GRCh38

View Full Product Details