Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32311727-32311988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRAP
Entrez Gene ID
56246
Gene Name
melanocortin 2 receptor accessory protein
Gene Aliases
B27, C21orf61, FALP, FGD2, GCCD2
UniGene
Hs.584940
Gene Chromosome Location
Chr.21: 32291813-32314784 on Build GRCh38
Transcripts
NM_001285394.1 NM_178817.3 NM_206898.1 XM_006724028.3 XM_017028407.1 NM_014825.2

Gene Symbol
URB1
Entrez Gene ID
9875
Gene Name
URB1 ribosome biogenesis 1 homolog (S. cerevisiae)
Gene Aliases
C21orf108, NPA1
UniGene
Hs.473611
Gene Chromosome Location
Chr.21: 32311019-32393003 on Build GRCh38
Transcripts
NM_001285394.1 NM_178817.3 NM_206898.1 XM_006724028.3 XM_017028407.1 NM_014825.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6160934 c.490G>A p.G164R Chr.21: 32311967-32311967 on Build GRCh38
368811 c.365C>A p.P122Q Chr.21: 32311842-32311842 on Build GRCh38
6038505 c.403G>T p.G135C Chr.21: 32311880-32311880 on Build GRCh38
5572082 c.432G>A p.W144* Chr.21: 32311909-32311909 on Build GRCh38
359736 c.486C>A p.P162P Chr.21: 32311963-32311963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748116900 Chr.21: 32311876-32311876 on Build GRCh38
rs777253425 Chr.21: 32311827-32311827 on Build GRCh38
rs755878119 Chr.21: 32311945-32311945 on Build GRCh38
rs748843176 Chr.21: 32311887-32311887 on Build GRCh38
rs201370496 Chr.21: 32311793-32311793 on Build GRCh38
rs781233445 Chr.21: 32311776-32311776 on Build GRCh38
rs766368326 Chr.21: 32311933-32311933 on Build GRCh38
rs766760000 Chr.21: 32311861-32311861 on Build GRCh38
rs769078228 Chr.21: 32311906-32311906 on Build GRCh38
rs765412733 Chr.21: 32311830-32311830 on Build GRCh38
rs146788457 Chr.21: 32311963-32311963 on Build GRCh38
rs745997524 Chr.21: 32311941-32311941 on Build GRCh38
rs377210171 Chr.21: 32311877-32311877 on Build GRCh38
rs139379303 Chr.21: 32311799-32311799 on Build GRCh38
rs1020937788 Chr.21: 32311859-32311859 on Build GRCh38
rs755085204 Chr.21: 32311937-32311937 on Build GRCh38
rs777725817 Chr.21: 32311954-32311954 on Build GRCh38
rs74913568 Chr.21: 32311761-32311761 on Build GRCh38
rs747287896 Chr.21: 32311751-32311752 on Build GRCh38
rs754268566 Chr.21: 32311758-32311758 on Build GRCh38
rs778228116 Chr.21: 32311956-32311956 on Build GRCh38
rs200921993 Chr.21: 32311836-32311836 on Build GRCh38
rs756817659 Chr.21: 32311955-32311955 on Build GRCh38
rs376465589 Chr.21: 32311748-32311748 on Build GRCh38
rs758718535 Chr.21: 32311847-32311847 on Build GRCh38
rs143558834 Chr.21: 32311959-32311959 on Build GRCh38
rs370691714 Chr.21: 32311774-32311774 on Build GRCh38
rs763168814 Chr.21: 32311931-32311931 on Build GRCh38
rs114530014 Chr.21: 32311866-32311866 on Build GRCh38
rs150866700 Chr.21: 32311778-32311778 on Build GRCh38
rs779122471 Chr.21: 32311762-32311762 on Build GRCh38
rs965318901 Chr.21: 32311860-32311860 on Build GRCh38
rs374356072 Chr.21: 32311801-32311801 on Build GRCh38
rs376924425 Chr.21: 32311942-32311942 on Build GRCh38
rs777077799 Chr.21: 32311965-32311965 on Build GRCh38
rs761106555 Chr.21: 32311753-32311753 on Build GRCh38
rs751870972 Chr.21: 32311936-32311936 on Build GRCh38
rs1035725794 Chr.21: 32311926-32311926 on Build GRCh38
rs1009755163 Chr.21: 32311848-32311848 on Build GRCh38
rs777284724 Chr.21: 32311882-32311882 on Build GRCh38
rs564594808 Chr.21: 32311951-32311951 on Build GRCh38
rs769302607 Chr.21: 32311780-32311780 on Build GRCh38
rs768317959 Chr.21: 32311775-32311775 on Build GRCh38
rs764709506 Chr.21: 32311755-32311755 on Build GRCh38
rs369795618 Chr.21: 32311901-32311901 on Build GRCh38
rs772639515 Chr.21: 32311782-32311782 on Build GRCh38
rs765792719 Chr.21: 32311927-32311927 on Build GRCh38
rs757872679 Chr.21: 32311844-32311845 on Build GRCh38
rs774119464 Chr.21: 32311899-32311899 on Build GRCh38
rs150008653 Chr.21: 32311843-32311843 on Build GRCh38
rs781756108 Chr.21: 32311931-32311931 on Build GRCh38
rs17855143 Chr.21: 32311867-32311867 on Build GRCh38
rs773835535 Chr.21: 32311928-32311928 on Build GRCh38
rs776794313 Chr.21: 32311910-32311910 on Build GRCh38
rs868744185 Chr.21: 32311834-32311836 on Build GRCh38
rs770396189 Chr.21: 32311894-32311894 on Build GRCh38
rs534626403 Chr.21: 32311763-32311763 on Build GRCh38
rs552729042 Chr.21: 32311800-32311800 on Build GRCh38
rs575054047 Chr.21: 32311864-32311864 on Build GRCh38
rs200448756 Chr.21: 32311923-32311923 on Build GRCh38
rs764361483 Chr.21: 32311809-32311809 on Build GRCh38
rs780029065 Chr.21: 32311770-32311770 on Build GRCh38

View Full Product Details