Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38731694-38731955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALM
Entrez Gene ID
130589
Gene Name
galactose mutarotase
Gene Aliases
BLOCK25, GLAT, HEL-S-63p, IBD1
UniGene
Hs.435012
Gene Chromosome Location
Chr.2: 38665910-38734767 on Build GRCh38
Transcripts
NM_138801.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3183912 c.786T>C p.H262H Chr.2: 38731744-38731744 on Build GRCh38
4094160 c.922A>G p.T308A Chr.2: 38731880-38731880 on Build GRCh38
6668951 c.888T>C p.A296A Chr.2: 38731846-38731846 on Build GRCh38
5651904 c.863G>A p.G288D Chr.2: 38731821-38731821 on Build GRCh38
3183916 c.846G>A p.T282T Chr.2: 38731804-38731804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527668111 Chr.2: 38731754-38731754 on Build GRCh38
rs751290479 Chr.2: 38731881-38731881 on Build GRCh38
rs377121126 Chr.2: 38731787-38731787 on Build GRCh38
rs200329326 Chr.2: 38731738-38731738 on Build GRCh38
rs1040600127 Chr.2: 38731847-38731847 on Build GRCh38
rs762686164 Chr.2: 38731866-38731866 on Build GRCh38
rs979530350 Chr.2: 38731810-38731810 on Build GRCh38
rs758153760 Chr.2: 38731775-38731775 on Build GRCh38
rs776913901 Chr.2: 38731845-38731845 on Build GRCh38
rs752210344 Chr.2: 38731916-38731916 on Build GRCh38
rs200427606 Chr.2: 38731746-38731746 on Build GRCh38
rs552741943 Chr.2: 38731758-38731758 on Build GRCh38
rs778282803 Chr.2: 38731720-38731720 on Build GRCh38
rs749835753 Chr.2: 38731724-38731724 on Build GRCh38
rs750187952 Chr.2: 38731772-38731772 on Build GRCh38
rs1034886694 Chr.2: 38731784-38731784 on Build GRCh38
rs887870882 Chr.2: 38731761-38731761 on Build GRCh38
rs750905462 Chr.2: 38731926-38731926 on Build GRCh38
rs746235250 Chr.2: 38731807-38731807 on Build GRCh38
rs766236363 Chr.2: 38731873-38731873 on Build GRCh38
rs146302155 Chr.2: 38731865-38731865 on Build GRCh38
rs772191552 Chr.2: 38731744-38731744 on Build GRCh38
rs929483358 Chr.2: 38731878-38731878 on Build GRCh38
rs780730472 Chr.2: 38731788-38731788 on Build GRCh38
rs200689749 Chr.2: 38731888-38731888 on Build GRCh38
rs761914535 Chr.2: 38731849-38731849 on Build GRCh38
rs545817619 Chr.2: 38731804-38731804 on Build GRCh38
rs148753204 Chr.2: 38731841-38731841 on Build GRCh38
rs369470594 Chr.2: 38731753-38731753 on Build GRCh38
rs1022943042 Chr.2: 38731742-38731742 on Build GRCh38
rs774638353 Chr.2: 38731734-38731734 on Build GRCh38
rs553372506 Chr.2: 38731860-38731860 on Build GRCh38
rs767153310 Chr.2: 38731890-38731890 on Build GRCh38
rs1047863001 Chr.2: 38731880-38731880 on Build GRCh38
rs775609819 Chr.2: 38731820-38731820 on Build GRCh38
rs201512409 Chr.2: 38731825-38731825 on Build GRCh38
rs536111413 Chr.2: 38731844-38731844 on Build GRCh38
rs570948999 Chr.2: 38731770-38731770 on Build GRCh38
rs564142030 Chr.2: 38731864-38731864 on Build GRCh38
rs771406146 Chr.2: 38731727-38731727 on Build GRCh38
rs149331676 Chr.2: 38731803-38731803 on Build GRCh38
rs370570697 Chr.2: 38731800-38731800 on Build GRCh38
rs114039092 Chr.2: 38731813-38731813 on Build GRCh38
rs900477685 Chr.2: 38731811-38731811 on Build GRCh38
rs373233987 Chr.2: 38731786-38731786 on Build GRCh38
rs943657006 Chr.2: 38731842-38731842 on Build GRCh38
rs755566535 Chr.2: 38731919-38731919 on Build GRCh38
rs765969103 Chr.2: 38731776-38731776 on Build GRCh38

View Full Product Details