Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92455505-92455744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDE
Entrez Gene ID
3416
Gene Name
insulin degrading enzyme
Gene Aliases
INSULYSIN
UniGene
Hs.500546
Gene Chromosome Location
Chr.10: 92451684-92574484 on Build GRCh38
Transcripts
NM_001165946.1 NM_001322793.1 NM_001322794.1 NM_001322795.1 NM_001322796.1 NM_001322797.1 NM_004969.3 NR_136399.1 XM_017016187.1 XM_017016188.1 XM_017016189.1 XM_017016190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7182262 c.2930A>G p.D977G Chr.10: 92455610-92455610 on Build GRCh38
4016624 c.2935A>C p.N979H Chr.10: 92455605-92455605 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs188614842 Chr.10: 92455596-92455596 on Build GRCh38
rs200909421 Chr.10: 92455673-92455673 on Build GRCh38
rs374799415 Chr.10: 92455561-92455561 on Build GRCh38
rs184522345 Chr.10: 92455566-92455566 on Build GRCh38
rs764357071 Chr.10: 92455692-92455693 on Build GRCh38
rs776701450 Chr.10: 92455631-92455631 on Build GRCh38
rs942069161 Chr.10: 92455565-92455565 on Build GRCh38
rs772369342 Chr.10: 92455599-92455599 on Build GRCh38
rs964969638 Chr.10: 92455711-92455711 on Build GRCh38
rs1013758033 Chr.10: 92455655-92455655 on Build GRCh38
rs11459510 Chr.10: 92455707-92455708 on Build GRCh38
rs761147297 Chr.10: 92455609-92455609 on Build GRCh38
rs182658923 Chr.10: 92455700-92455700 on Build GRCh38
rs765469356 Chr.10: 92455633-92455633 on Build GRCh38
rs1031638404 Chr.10: 92455591-92455591 on Build GRCh38
rs201349668 Chr.10: 92455608-92455608 on Build GRCh38
rs751353310 Chr.10: 92455680-92455680 on Build GRCh38
rs397768997 Chr.10: 92455712-92455713 on Build GRCh38
rs746254271 Chr.10: 92455588-92455588 on Build GRCh38
rs896299375 Chr.10: 92455646-92455646 on Build GRCh38
rs763200714 Chr.10: 92455660-92455660 on Build GRCh38
rs780624910 Chr.10: 92455557-92455557 on Build GRCh38
rs528166237 Chr.10: 92455703-92455703 on Build GRCh38
rs761845335 Chr.10: 92455632-92455632 on Build GRCh38
rs750696259 Chr.10: 92455638-92455638 on Build GRCh38
rs769153466 Chr.10: 92455623-92455623 on Build GRCh38
rs978823534 Chr.10: 92455576-92455576 on Build GRCh38
rs917106778 Chr.10: 92455649-92455649 on Build GRCh38
rs751489134 Chr.10: 92455542-92455542 on Build GRCh38
rs780078518 Chr.10: 92455562-92455562 on Build GRCh38
rs796803608 Chr.10: 92455700-92455701 on Build GRCh38
rs754937274 Chr.10: 92455553-92455553 on Build GRCh38
rs149732626 Chr.10: 92455694-92455694 on Build GRCh38
rs573196074 Chr.10: 92455702-92455703 on Build GRCh38
rs992717292 Chr.10: 92455639-92455639 on Build GRCh38
rs779878573 Chr.10: 92455535-92455535 on Build GRCh38
rs919868311 Chr.10: 92455580-92455580 on Build GRCh38

View Full Product Details