Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 17816593-17816818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSGN1
Entrez Gene ID
343930
Gene Name
mesogenin 1
Gene Aliases
MSOG
UniGene
Hs.705359
Gene Chromosome Location
Chr.2: 17816519-17817100 on Build GRCh38
Transcripts
NM_001105569.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3896113 c.180C>T p.P60P Chr.2: 17816698-17816698 on Build GRCh38
3896114 c.300C>T p.F100F Chr.2: 17816818-17816818 on Build GRCh38
3933483 c.200T>G p.L67R Chr.2: 17816718-17816718 on Build GRCh38
6089162 c.202C>T p.P68S Chr.2: 17816720-17816720 on Build GRCh38
359403 c.180C>A p.P60P Chr.2: 17816698-17816698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368478996 Chr.2: 17816686-17816688 on Build GRCh38
rs772451330 Chr.2: 17816674-17816674 on Build GRCh38
rs749887278 Chr.2: 17816779-17816779 on Build GRCh38
rs879213251 Chr.2: 17816739-17816739 on Build GRCh38
rs753107164 Chr.2: 17816613-17816613 on Build GRCh38
rs748059839 Chr.2: 17816741-17816741 on Build GRCh38
rs780978531 Chr.2: 17816737-17816737 on Build GRCh38
rs758007424 Chr.2: 17816729-17816729 on Build GRCh38
rs375175859 Chr.2: 17816677-17816677 on Build GRCh38
rs796209036 Chr.2: 17816653-17816653 on Build GRCh38
rs769857552 Chr.2: 17816742-17816742 on Build GRCh38
rs779759038 Chr.2: 17816640-17816640 on Build GRCh38
rs762244506 Chr.2: 17816766-17816766 on Build GRCh38
rs149704294 Chr.2: 17816776-17816776 on Build GRCh38
rs758062614 Chr.2: 17816638-17816638 on Build GRCh38
rs1048970746 Chr.2: 17816777-17816777 on Build GRCh38
rs779093390 Chr.2: 17816745-17816745 on Build GRCh38
rs769125053 Chr.2: 17816763-17816763 on Build GRCh38
rs756836340 Chr.2: 17816628-17816628 on Build GRCh38
rs200591173 Chr.2: 17816760-17816760 on Build GRCh38
rs912920270 Chr.2: 17816751-17816751 on Build GRCh38
rs759764123 Chr.2: 17816670-17816670 on Build GRCh38
rs928234552 Chr.2: 17816615-17816615 on Build GRCh38
rs369675155 Chr.2: 17816701-17816701 on Build GRCh38
rs764480289 Chr.2: 17816700-17816700 on Build GRCh38
rs779703784 Chr.2: 17816730-17816730 on Build GRCh38
rs139339372 Chr.2: 17816704-17816704 on Build GRCh38
rs745963480 Chr.2: 17816747-17816747 on Build GRCh38
rs749940776 Chr.2: 17816720-17816720 on Build GRCh38
rs745581168 Chr.2: 17816632-17816632 on Build GRCh38
rs768428754 Chr.2: 17816647-17816647 on Build GRCh38
rs749323702 Chr.2: 17816655-17816655 on Build GRCh38
rs553482765 Chr.2: 17816759-17816759 on Build GRCh38
rs776876631 Chr.2: 17816765-17816765 on Build GRCh38
rs772398177 Chr.2: 17816755-17816755 on Build GRCh38
rs946759828 Chr.2: 17816708-17816708 on Build GRCh38
rs774585399 Chr.2: 17816665-17816665 on Build GRCh38
rs753262043 Chr.2: 17816627-17816627 on Build GRCh38
rs771193106 Chr.2: 17816657-17816657 on Build GRCh38
rs558058990 Chr.2: 17816694-17816694 on Build GRCh38
rs34069439 Chr.2: 17816758-17816758 on Build GRCh38
rs774376089 Chr.2: 17816643-17816643 on Build GRCh38
rs1040261976 Chr.2: 17816771-17816771 on Build GRCh38
rs201149255 Chr.2: 17816734-17816734 on Build GRCh38
rs746853787 Chr.2: 17816643-17816643 on Build GRCh38
rs778536349 Chr.2: 17816630-17816630 on Build GRCh38
rs756535632 Chr.2: 17816620-17816620 on Build GRCh38

View Full Product Details